Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Pricing and availability is not currently available.
Supelco
Melting point standard 121-123°C, analytical standard
Pricing and availability is not currently available.
Supelco
4-Hydroxybenzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Syringaldehyde, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Benzoic acid, SAJ first grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Benzoic acid, SAJ special grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Pricing and availability is not currently available.
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Pricing and availability is not currently available.
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
trans-Sinapic acid, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, for chromatography
Pricing and availability is not currently available.
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
trans-p-Coumaric acid, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Pricing and availability is not currently available.
Supelco
Propionic acid, analytical standard
Pricing and availability is not currently available.
Supelco
trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Pricing and availability is not currently available.
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Methoxyphenylacetic acid, ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Hydroxycinnamic acid, predominantly trans, 97%
Pricing and availability is not currently available.