Merck
  • A charge neutral, size tuneable polymersome capable of high biological encapsulation efficiency and cell permeation.
A charge neutral, size tuneable polymersome capable of high biological encapsulation efficiency and cell permeation.
International journal of pharmaceutics (2015-01-17)
Chloe Martin, Elsa Dolmazon, Katrina Moylan, Colin Fowley, Anthony P McHale, John F Callan, Bridgeen Callan
ABSTRACT
The field of therapeutics is evolving to include a greater proportion of higher molecular weight, hydrophilic biological compounds. To cater for this new era in healthcare the concomitant development of appropriate drug delivery systems is essential to aid cellular permeation. In this manuscript we present the synthesis, characterisation and biological evaluation of a charge neutral polymersome (Ps) based drug delivery system (DDS) using an amphiphilic pegylated random copolymer. A detailed dynamic light scattering study revealed that the hydrodynamic diameter of the Ps can be tailored to a specific size simply by varying the quantities and ratios used during the preparation step. The zeta potential of this new drug delivery system was determined to be -0.095 ± 0.037 mV, the encapsulation efficiency of Fitc-CM-Dextran (4 KDa) was 70%, the uptake of Fitc-CM-Dextran by Hela cells was increased 4-fold when encapsulated within the polymersomal system. The facile preparation, high loading capacity and size tuneable nature of this Ps renders it a promising alternative to the ever growing array of currently available Ps.
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
1-Decanol, ≥98%, FCC, FG
Sigma-Aldrich
Methacrylic acid, contains 250 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
1-Decanol, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Methacrylic acid, SAJ first grade, ≥98.0%
Sigma-Aldrich
FITC-CM-Dextran, average mol wt 70,000
Sigma-Aldrich
FITC-CM-Dextran, average mol wt 150,000
Sigma-Aldrich
FITC-CM-Dextran, average mol wt 4,000
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Supelco
1-Decanol, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
1-Decanol, 98%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard