Merck
  • NLRP7 and KHDC3L, the two maternal-effect proteins responsible for recurrent hydatidiform moles, co-localize to the oocyte cytoskeleton.
NLRP7 and KHDC3L, the two maternal-effect proteins responsible for recurrent hydatidiform moles, co-localize to the oocyte cytoskeleton.
Human reproduction (Oxford, England) (2014-11-02)
Elie Akoury, Li Zhang, Asangla Ao, Rima Slim
ABSTRACT
What is the subcellular localization in human oocytes and preimplantation embryos, of the two maternal-effect proteins, NLRP7 and KHDC3L, responsible for recurrent hydatidiform moles (RHMs)? NLRP7 and KHDC3L localize to the oocyte cytoskeleton and are polar and absent from the cell-to-cell contact region in early preimplantation embryos. NLRP7 and KHDC3L expression has been described at the RNA level in some stages of human oocytes and preimplantation embryos and at the protein level by immunohistochemistry in human and bovine ovaries. NLRP7 and KHDC3L co-localize to the microtubule organizing center and/or the Golgi apparatus in human hematopoietic cells. A total of 164 spare human oocytes and embryos from patients undergoing in vitro fertilization were used. Oocytes and early cleavage-stage embryos were fixed, immunostained with NLRP7 and/or KHDC3L antibodies, and analyzed using high-resolution confocal immunofluorescence and electron microscopies. NLRP7 and KHDC3L localize to the cytoskeleton and are predominant at the cortical region in growing oocytes. After the first cellular division, these two maternal-effect proteins become asymmetrically confined to the outer cortical region and excluded from the cell-to-cell contact region until the blastocyst stage where NLRP7 and KHDC3L homogeneously redistribute to the cytoplasm and the nucleus, respectively. We could not analyze fresh human oocytes and embryos. The analyzed materials were donated by patients undergoing assisted reproductive technologies and released for research 1-3 days after their collection and the transfer of embryos to the patients. Our study is the first comprehensive and high-resolution localization of the only two known maternal-effect proteins, NLRP7 and KHDC3L, in human oocytes and preimplantation embryos. Our data contribute to a better understanding of the roles of these two proteins in the integrity of the oocytes, post-zygotic divisions, and cell-lineage differentiation. This work was supported by the Canadian Institute of Health Research (86546 to R.S.); E.A. was supported by fellowships from the Research Institute of the McGill University Health Centre and a CREATE award from the Réseau Québécois en Reproduction. All authors declare no conflict of interest.
Product Description
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
Sigma-Aldrich
Glutaraldehyde solution, SAJ first grade, 20.0-26.0%
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–Gold antibody produced in rabbit, affinity isolated antibody, aqueous glycerol suspension, 10 nm (colloidal gold)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Glycine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications