Merck
  • Gas chromatography-mass spectrometry method optimized using response surface modeling for the quantitation of fungal off-flavors in grapes and wine.
Gas chromatography-mass spectrometry method optimized using response surface modeling for the quantitation of fungal off-flavors in grapes and wine.
Journal of agricultural and food chemistry (2015-02-24)
Navideh Sadoughi, Leigh M Schmidtke, Guillaume Antalick, John W Blackman, Christopher C Steel
ABSTRACT
An optimized method for the quantitation of volatile compounds responsible for off-aromas, such as earthy odors, found in wine and grapes was developed. The method involved a fast and simple headspace solid-phase microextraction-gas chromatography-mass spectrometry (HS-SPME-GC-MS) for simultaneous determination of 2-isopropyl-3-methoxypyrazine, 2-isobutyl-3-methoxypyrazine, 3-octanone, fenchone, 1-octen-3-one, trans-2-octen-1-ol, fenchol, 1-octen-3-ol, 2-methylisoborneol, 2,4,6-trichloroanisole, geosmin, 2,4,6-tribromoanisole, and pentachloroanisole. The extraction of the temperature and time were optimized using response surface methodology in both wine base (WB) and grape base (GB). Low limits of detection (0.1-5 ng/L in WB and 0.05-1.6 in GB) and quantitation (0.3-17 in WB and 0.2-6.2 in GB) with good recoveries (83-131%) and repeatability [4.3-9.8% coefficient of variation (CV) in WB and 5.1-11.1% CV in GB] and reproducibility (3.6-10.2 in WB and 1.9-10.9 in GB) indicate that the method has excellent sensitivity and is suitable for the analysis of these off-flavor compounds in wine and grape juice samples.
Product Description
Supelco
(±)-2-Octanol, analytical standard
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
L-(+)-Tartaric acid, SAJ first grade, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
L-(+)-Tartaric acid, JIS special grade, ≥99.5%
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME StableFlex fiber assortment kit, for use with manual holder
Supelco
SPME fiber assortment kit 1, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, size 24 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 23 ga, StableFlex fiber, for use with autosampler
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 23 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assortment kit 2, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 2, for use with autosampler, size 24 ga
Supelco
SPME fiber assortment kit 4, for use with manual holder, size 24 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with autosampler
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
L-(+)-Tartaric acid, ≥99.5%
Supelco
L-(+)-Tartaric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
1-Octen-3-ol, 98%
Sigma-Aldrich
1-Octen-3-ol, ≥98%, FCC, FG
Sigma-Aldrich
L-(+)-Tartaric acid, puriss. p.a., reag. ISO, 99.5-101.0% (calc. to the dried substance)