Merck
Hypoxic potentiation of nitrite effects in human vessels and platelets.
Nitric oxide : biology and chemistry (2014-05-27)
Rustem F Dautov, Irene Stafford, Saifei Liu, Hugh Cullen, M Madhani, Yuliy Y Chirkov, John D Horowitz
ABSTRACT
Previous studies in non-human blood vessels and in platelets have demonstrated that under hypoxic conditions release of NO from nitrite (NO2(-)) is potentiated by deoxyhaemoglobin. In the current study, we characterized hypoxic potentiation of NO2(-) effects in human vasculature and platelets in vitro, addressing underlying mechanisms. The vasodilator efficacy of NO2(-), in comparison with glyceryl trinitrate (GTN), was evaluated in vitro, using segments of human saphenous vein. Under hypoxic conditions, there was a leftward shift of the NO2(-) concentration-response curve (EC50: 22 μM in hyperoxia vs 3.5 μM in hypoxia; p<0.01), but no significant potentiation of GTN effect. In the presence of red blood cells, hypoxic potentiation of NO2(-) vasodilator effect was accentuated. In whole blood samples and platelet-rich plasma (PRP) we assessed inhibition of platelet aggregation by NO2(-) (1mM), in comparison with that of sodium nitroprusside (SNP, 10 μM). In individual subjects (n=37), there was a strong correlation (r=0.75, p<0.0001) between anti-aggregatory effects of NO2(-) and SNP in whole blood, signifying that resultant sGC activation underlies biological effect and responses to NO2(-) are diminished in the presence of NO resistance. In PRP, the effects of NO2(-) were less pronounced than in whole blood (p=0.0001), suggesting an important role of Hb (within RBCs) in the bioconversion of NO2(-) to NO. Inhibition of platelet aggregation by NO2(-) was almost 3-fold greater in venous than in arterial blood (p<0.0001), and deoxyHb concentration directly correlated (r=0.69, p=0.013) with anti-aggregatory response. Incremental hypoxia applied to venous blood samples (in hypoxic chamber) caused a progressive increase in both deoxyHb level and anti-aggregatory effect of NO2(-). When subjects inhaled a 12% O2 mixture for 20 min, there was a 3-fold rise in blood deoxyHb fraction (p<0.01). In PRP, response to NO2(-) also increased under hypoxia, and was further enhanced (p<0.01) by deoxyHb. Furthermore, deoxyHb exerted significant anti-aggregatory effects even in the absence of added NO2(-), suggesting a role for endogenous NO2(-). The results of this work provide further mechanistic insights into hypoxic potentiation of vasodilator and anti-aggregatory actions of NO2(-). In human saphenous veins and blood, the balance of evidence suggests differential rates of NO release from NO2(-) (largely modulated by deoxyHb) as the fundamental mechanism.
Product Description
Supelco
Nitrite ion standard solution, 0.1 M NO2-, for ion-selective electrodes
Sigma-Aldrich
Adenosine 5′-diphosphate, ≥95% (HPLC)
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Supelco
Sodium nitrite, analytical standard
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
Supelco
Sodium Nitroprusside, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%