Merck
  • Comparison of the antifungal efficacy of terbinafine hydrochloride and ciclopirox olamine containing formulations against the dermatophyte Trichophyton rubrum in an infected nail plate model.
Comparison of the antifungal efficacy of terbinafine hydrochloride and ciclopirox olamine containing formulations against the dermatophyte Trichophyton rubrum in an infected nail plate model.
Molecular pharmaceutics (2014-02-05)
Anja Täuber, Christel C Müller-Goymann
ABSTRACT
Onychomycosis is a fungal infection mostly induced by dermatophytes such as Trichophyton rubrum. Due to slow nail growth, the treatment takes 3-9 months depending on the nail size and infected area. Hence, high efficacy of the active ingredient without systemic side effects is of major interest. To test the efficacy of an antifungal formulation, an appropriate in vitro model reflecting the in vivo situation as close as possible is required. In this study, a variety of antifungal formulations, i.e., commercial ones (Ciclopoli and Lamisil cream), those used in compounding pharmacies (Pentravan) as well as poloxamer 407-based systems, have been evaluated in an infected nail plate model. The active pharmaceutical ingredients (APIs) were ciclopirox olamine and terbinafine hydrochloride. The poloxamer 407-based formulations consisted of poloxamer 407, double distilled water, propylene glycol, isopropyl alcohol, medium chain triglycerides and either 1% ciclopirox olamine or 1% terbinafine hydrochloride as API, respectively. Former studies have shown high permeation rates of terbinafine hydrochloride from similar poloxamer 407-based formulations with dimethyl isosorbide instead of propylene glycol. The present contribution shows superior inhibition of T. rubrum growth from poloxamer 407-based formulations in comparison to the commercial Lamisil cream. Moreover, poloxamer 407-based formulations were equally effective as the nail lacquer Ciclopoli even though the poloxamer formulations contained only 1% of the drug instead of 8% in the marketed lacquer. Poloxamer 407-based systems containing ciclopirox olamine proved to be about as effective as similar terbinafine hydrochloride systems.
Sigma-Aldrich
2-Mercaptoethanol, SAJ special grade, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Thiourea, JIS special grade, ≥98.0%
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, BioXtra, ≥98.0% (T)
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, tested according to Ph. Eur.
Millipore
Urea solution, suitable for microbiology, 40% in H2O