Merck
  • Constitutive expression of a COOH-terminal leucine mutant of lysosome-associated membrane protein-1 causes its exclusive localization in low density intracellular vesicles.
Constitutive expression of a COOH-terminal leucine mutant of lysosome-associated membrane protein-1 causes its exclusive localization in low density intracellular vesicles.
Journal of biochemistry (2014-04-04)
Kenji Akasaki, Keiko Shiotsu, Akihiro Michihara, Norie Ide, Ikuo Wada
ABSTRACT
Lysosome-associated membrane protein-1 (LAMP-1) is a type I transmembrane protein with a short cytoplasmic tail that possesses a lysosome-targeting signal of GYQTI(382)-COOH. Wild-type (WT)-LAMP-1 was exclusively localized in high density lysosomes, and efficiency of LAMP-1's transport to lysosomes depends on its COOH-terminal amino acid residue. Among many different COOH-terminal amino acid substitution mutants of LAMP-1, a leucine-substituted mutant (I382L) displays the most efficient targeting to late endosomes and lysosomes [Akasaki et al. (2010) J. Biochem. 148: , 669-679]. In this study, we generated two human hepatoma cell lines (HepG2 cell lines) that stably express WT-LAMP-1 and I382L, and compared their intracellular distributions. The subcellular fractionation study using Percoll density gradient centrifugation revealed that WT-LAMP-1 had preferential localization in the high density secondary lysosomes where endogenous human LAMP-1 was enriched. In contrast, a major portion of I382L was located in a low density fraction. The low density fraction also contained approximately 80% of endogenous human LAMP-1 and significant amounts of endogenous β-glucuronidase and LAMP-2, which probably represents occurrence of low density lysosomes in the I382L-expressing cells. Double immunofluorescence microscopic analyses distinguished I382L-containing intracellular vesicles from endogenous LAMP-1-containing lysosomes and early endosomes. Altogether, constitutive expression of I382L causes its aberrant intracellular localization and generation of low density lysosomes, indicating that the COOH-terminal isoleucine is critical for normal localization of LAMP-1 in the dense lysosomes.
Product Description
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
SAFC
L-Glutamine