Merck
  • Probing of a human proteome microarray with a recombinant pathogen protein reveals a novel mechanism by which hookworms suppress B-cell receptor signaling.
Probing of a human proteome microarray with a recombinant pathogen protein reveals a novel mechanism by which hookworms suppress B-cell receptor signaling.
The Journal of infectious diseases (2014-08-21)
Leon Tribolet, Cinzia Cantacessi, Darren A Pickering, Severine Navarro, Denise L Doolan, Angela Trieu, Huang Fei, Yang Chao, Andreas Hofmann, Robin B Gasser, Paul R Giacomin, Alex Loukas
ABSTRACT
Na-ASP-2 is an efficacious hookworm vaccine antigen. However, despite elucidation of its crystal structure and studies addressing its immunobiology, the function of Na-ASP-2 has remained elusive. We probed a 9000-protein human proteome microarray with Na-ASP-2 and showed binding to CD79A, a component of the B-cell antigen receptor complex. Na-ASP-2 bound to human B lymphocytes ex vivo and downregulated the transcription of approximately 1000 B-cell messenger RNAs (mRNAs), while only approximately 100 mRNAs were upregulated, compared with control-treated cells. The expression of a range of molecules was affected by Na-ASP-2, including factors involved in leukocyte transendothelial migration pathways and the B-cell signaling receptor pathway. Of note was the downregulated transcription of lyn and pi3k, molecules that are known to interact with CD79A and control B-cell receptor signaling processes. Together, these results highlight a previously unknown interaction between a hookworm-secreted protein and B cells, which has implications for helminth-driven immunomodulation and vaccine development. Further, the novel use of human protein microarrays to identify host-pathogen interactions, coupled with ex vivo binding studies and subsequent analyses of global gene expression in human host cells, demonstrates a new pipeline by which to explore the molecular basis of infectious diseases.
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, SAJ special grade, ≥99.0%