Merck
  • Chronic treatment with novel brain-penetrating selective NOP receptor agonist MT-7716 reduces alcohol drinking and seeking in the rat.
Chronic treatment with novel brain-penetrating selective NOP receptor agonist MT-7716 reduces alcohol drinking and seeking in the rat.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology (2014-05-28)
Roberto Ciccocioppo, Serena Stopponi, Daina Economidou, Makoto Kuriyama, Hiroshi Kinoshita, Markus Heilig, Marisa Roberto, Friedbert Weiss, Koji Teshima
ABSTRACT
Since its discovery, the nociceptin/orphanin FQ (N/OFQ)-NOP receptor system has been extensively investigated as a promising target to treat alcoholism. Encouraging results obtained with the endogenous ligand N/OFQ stimulated research towards the development of novel brain-penetrating NOP receptor agonists with a pharmacological and toxicological profile compatible with clinical development. Here we describe the biochemical and alcohol-related behavioral effects of the novel NOP receptor agonist MT-7716. MT-7716 has high affinity for human NOP receptors expressed in HEK293 cells with a Ki value of 0.21 nM. MT-7716 concentration-dependently stimulated GTPγ(35)S binding with an EC50 value of 0.30 nM and its efficacy was similar to N/OFQ, suggesting that MT7716 is a full agonist at NOP receptors. In the two bottle choice test MT-7716 (0, 0.3, 1, and 3 mg/kg, bid) given orally for 14 days dose-dependently decreased voluntary alcohol intake in Marchigian Sardinian rats. The effect became gradually stronger following repeated administration, and was still significant 1 week after discontinuation of the drug. Oral naltrexone (30 mg/kg, bid) for 14 days also reduced ethanol intake; however, the effect decreased over the treatment period and rapidly disappeared when drug treatment was discontinued. MT-7716 is also effective for preventing reinstatement caused by both ethanol-associated environmental stimuli and stress. Finally, to investigate the effect of MT-7716 on alcohol withdrawal symptoms, Wistar rats were withdrawn from a 7-day alcohol liquid diet. MT-7716 significantly attenuated somatic alcohol withdrawal symptoms. Together these findings indicate that MT-7716 is a promising candidate for alcoholism treatment remaining effective with chronic administration.
Sigma-Aldrich
Naltrexone hydrochloride
Sigma-Aldrich
Yohimbine hydrochloride, ≥98% (HPLC), powder
Yohimbine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Naltrexone hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acamprosate calcium, ≥98% (HPLC), powder