Merck
  • Antimicrobial activity of ibuprofen: new perspectives on an "Old" non-antibiotic drug.
Antimicrobial activity of ibuprofen: new perspectives on an "Old" non-antibiotic drug.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2015-02-25)
Jelena Obad, Jagoda Šušković, Blaženka Kos
ABSTRACT
Pharmaceutical industry has been encountering antimicrobial activity of non-antibiotics during suitability tests carried out prior to routine pharmacopoeial microbiological purity analysis of finished dosage forms. These properties are usually ignored or perceived as a nuisance during pharmaceutical analysis. The aim of this study was: (i) to compare the available data to our method suitability test results carried out on products containing ibuprofen, i.e. to demonstrate that method suitability can be a valuable tool in identifying new antimicrobials, (ii) to demonstrate the antimicrobial activity of ibuprofen and ibuprofen lysine. Microbiological purity method suitability testing was carried out according to European Pharmacopoeia (EP), chapters 2.6.12. and 2.6.13. Antimicrobial activity of ibuprofen and ibuprofen lysine was demonstrated by a disk diffusion method, a modification of the European Committee for Antimicrobial Susceptibility Testing method (EUCAST), against test microorganisms recommended in the EP. It was confirmed that ibuprofen may be responsible for the broad spectrum of antimicrobial activity of the tested products, and that method suitability tests according to the EP can indeed be exploited by the scientific community in setting guidelines towards future research of new antimicrobials. In the disk diffusion assay, inhibition zones were obtained with more than 62.5 μg and 250 μg for Staphylococcus aureus, 125 μg and 250 μg for Bacillus subtilis, 31.3 μg and 125 μg for Candidaalbicans, 31.3 μg and 62.5 μg for Aspergillusbrasiliensis, of ibuprofen/disk, and ibuprofen lysine/disk, respectively. For Escherichiacoli, Pseudomonasaeruginosa and Salmonellatyphimurium inhibition zones were not obtained. Antimicrobial activity of ibuprofen is considered merely as a side effect, and it is not mentioned in the patient information leaflets of ibuprofen drugs. As such, for the patient, it could represent an advantage, but, it could also introduce additional risks during usage. Further microbiological, pharmacological and clinical trials are of great importance.
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Supelco
(±)-Miconazole nitrate salt, Pharmaceutical Secondary Standard; Certified Reference Material
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Ibuprofen
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
(±)-Miconazole nitrate salt, imidazole antibiotic
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
USP
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
6-Mercaptohexanoic acid, 90%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Supelco
Amoxicillin trihydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Supelco
Amoxicillin trihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Amoxicillin trihydrate, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)