Merck
  • Cardiomyocyte apoptosis contributes to pathogenesis of cirrhotic cardiomyopathy in bile duct-ligated mice.
Cardiomyocyte apoptosis contributes to pathogenesis of cirrhotic cardiomyopathy in bile duct-ligated mice.
Clinical science (London, England : 1979) (2014-04-10)
Soon Woo Nam, Hongqun Liu, Joe Z Wong, Annie Y Feng, Gavin Chu, Naeem Merchant, Samuel S Lee
ABSTRACT
Cirrhotic cardiomyopathy is defined as systolic and diastolic dysfunctions, electrophysiological changes and macroscopic structural changes. However, the underlying mechanisms of this syndrome remain unclear. A possible role of myocardial apoptosis in the pathogenesis has not been previously examined. We hypothesized that dysregulation of apoptotic signalling participates in cardiac dysfunction in the cirrhotic heart. Therefore, we evaluated apoptotic pathways in the hearts of mice with chronic BDL (bile duct ligation). A cirrhotic cardiomyopathy model was induced by BDL in mice. Left ventricular geometry and volumes were evaluated by MRI. Intrinsic and extrinsic apoptotic pathways were evaluated by immunohistochemical staining and Western blot analysis. Fas-mediated apoptosis was inhibited by in vivo administration of an anti-FasL (Fas ligand) monoclonal antibody, and subsequently cardiac contractility was measured in isolated cardiomyocytes. BDL-mice showed significantly more PARP [poly(ADP-ribose) polymerase] staining than sham controls (18.2±11.4 compared with 6.7±5.3; P<0.05). Fas protein expression and PARP cleavage were activated, whereas FLIP (Fas-associated death domain-like interleukin 1β-converting enzyme-inhibitory protein) was decreased compared with sham controls. The Bcl-2/Bax ratio was increased in BDL-mice compared with sham controls. Anti-FasL monoclonal antibody injection in BDL-mice improved systolic and diastolic dysfunctions in cardiomyocytes, but had no effect in sham controls. A net pro-apoptotic balance exists in BDL hearts, mainly mediated by activation of the extrinsic pathway, and abrogation of apoptosis improved contractility. These results suggest that apoptosis contributes to depressed cardiac contractility in a murine model of cirrhotic cardiomyopathy.
Sigma-Aldrich
Rat Fas Ligand ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human Fas Ligand ELISA Kit, for serum, plasma, and cell culture supernatants
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Mouse Fas ELISA Kit
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)