Merck
  • Anti-anxiety self-medication in rats: oral consumption of chlordiazepoxide and ethanol after reward devaluation.
Anti-anxiety self-medication in rats: oral consumption of chlordiazepoxide and ethanol after reward devaluation.
Behavioural brain research (2014-09-23)
Lidia Manzo, Rocío Donaire, Marta Sabariego, Mauricio R Papini, Carmen Torres
ABSTRACT
Rats increased preference for ethanol after sessions of appetitive extinction, but not after acquisition (reinforced) sessions (Manzo et al., 2014). Drinking was not influenced by appetitive extinction in control groups with postsession access to water, rather than ethanol. Because ethanol has anxiolytic properties in tasks involving reward loss, these results were interpreted as anti-anxiety self-medication. The present experiment tested the potential for self-medication with the prescription anxiolytic chlordiazepoxide, a benzodiazepine with an addictive profile used in the treatment of anxiety disorders. To test this hypothesis, Wistar rats exposed to a 32-to-4% sucrose devaluation received a two-bottle, 2-h preference test immediately after consummatory training. One bottle contained 1 mg/kg of chlordiazepoxide, 2% ethanol, or water for different groups (the second bottle contained water for all groups). Three additional groups received the same postsession preference tests, but were exposed to 4% sucrose during consummatory training. Rats showed suppression of consummatory behavior after reward devaluation relative to unshifted controls. This effect was accompanied by a selective increase in preference for chlordiazepoxide and ethanol. Downshifted animals with access to water or unshifted controls with access to the anxiolytics failed to exhibit postsession changes in preference. Similar results were observed in terms of absolute consumption and consumption relative to body weight. This study shows for the first time that a prescription anxiolytic supports enhanced voluntary consumption during periods of emotional distress triggered by reward loss. Such anti-anxiety self-medication provides insights into the early stages of addictive behavior.
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Chlordiazepoxide hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology