Merck
  • Quantitative liquid chromatography-electrospray ionization-mass spectrometry analysis of amine-containing metabolites derivatized with cyanuric chloride and methylamine isotopologues.
Quantitative liquid chromatography-electrospray ionization-mass spectrometry analysis of amine-containing metabolites derivatized with cyanuric chloride and methylamine isotopologues.
Journal of chromatography. A (2015-03-04)
Der-Yen Lee, Geen-Dong Chang
ABSTRACT
Electrospray ionization (ESI) is the most useful interface for mass spectrometry associated with liquid chromatography (LC). However, analyses of polar metabolites become a challenge because the high polarity impairs the separation of metabolites in reversed-phase liquid chromatography and ionization of metabolites by ESI. In this article, we have used cyanuric chloride to couple the amine-containing molecules with methylamine and found that both resolution on LC chromatogram and ionization by ESI are greatly improved. Derivatives would be obtained in 2-h coupling reactions, and then resolved by LC-ESI-MS in 15 min for each sample. Most amino acids can be quantified with linear range from 1 nM to 1 μM and with an R(2) above 0.979. Although reversed-phase chromatography is suitable for resolving the derivatives, phenyl columns with methanol elution provide optimal separation and signal intensity. Moreover, most structural isomers are well separated following cyanuric chloride and methylamine derivatization. Instead of synthesizing a stable isotope-labeled cyanuric chloride, we can take advantage of using commercially available methyl-d3-amine for a novel stable isotope-coded derivatization method. Each metabolite can be directly quantified by the peak intensity ratio of each derivative isotopologue pair in a single LC-MS analysis. The coupling reactions are relatively easy and accessible to most investigators to generate multiple stable isotope-labeled derivatives of amine-containing compounds for a differential metabolomic analysis.
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
Methyl-d3-amine hydrochloride, 99 atom % D
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Supelco
L-Lysine, analytical standard
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Methylamine hydrochloride, ≥98%
Sigma-Aldrich
Cyanuric chloride, 99%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%