Merck
  • A multi-residue method for determination of 70 organic micropollutants in surface waters by solid-phase extraction followed by gas chromatography coupled to tandem mass spectrometry.
A multi-residue method for determination of 70 organic micropollutants in surface waters by solid-phase extraction followed by gas chromatography coupled to tandem mass spectrometry.
Environmental science and pollution research international (2014-08-12)
Evangelia Terzopoulou, Dimitra Voutsa, George Kaklamanos
ABSTRACT
A multi-residue method, based on gas chromatography coupled to tandem mass spectrometry (GC-MS/MS), has been developed for the determination of 70 organic micropollutants from various chemical classes (organochlorinated, organophosphorous, triazines, carbamate and urea, polycyclic aromatic hydrocarbons, polychlorinated biphenyls, pharmaceuticals, phenols, etc.) in surface waters. A single-step SPE extraction using OASIS HLB cartridges was employed for the recovery of target micropollutants. The method has been validated according to monitoring performance criteria of the Water Framework Directive, taking into account the approved guidelines on quality assurance and quality control. The recoveries ranged from 60 to 110 %, the coefficient of variation from 0.84 to 27.4 %, and the uncertainty from 6 to 37 %. The LOD varied from 6.0 to 40 ng/L. The limits of quantification for the priority pollutants anthracene, alachlor, atrazine, benzo(a)pyrene, chlorfenvinphos, diuron, isoproturon, nonylphenol, simazine, and terbutryn fulfill the criterion of <30 % of the relevant environmental standards. The method was employed to investigate the water quality in the basin of a transboundary river, Strymonas, in NE Greece during three sampling campaigns conducted in the year 2013. Thirty-nine compounds were detected in the river water. Metolachlor, diuron, isoproturon, salicylic acid, chlorfenvinphos, 1,2-benzanthracene, pyrene, diflubenzuron, and carbaryl exhibited the highest detection frequencies.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
2-Nitrophenol, 98%
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
USP
Salicylic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, SAJ special grade, ≥98.5%
Supelco
Bisphenol A, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
Benzotriazole, ReagentPlus®, 99%
Sigma-Aldrich
Bisphenol A, 97%
Sigma-Aldrich
Nonylphenol, technical grade, mixture of ring and chain isomers
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg