Merck
  • Neuronal driven pre-plaque inflammation in a transgenic rat model of Alzheimer's disease.
Neuronal driven pre-plaque inflammation in a transgenic rat model of Alzheimer's disease.
Neurobiology of aging (2014-05-17)
Cecilia E Hanzel, Alexa Pichet-Binette, Luisa S B Pimentel, M Florencia Iulita, Simon Allard, Adriana Ducatenzeiler, Sonia Do Carmo, A Claudio Cuello
ABSTRACT
Chronic brain inflammation is associated with Alzheimer's disease (AD) and is classically attributed to amyloid plaque deposition. However, whether the amyloid pathology can trigger early inflammatory processes before plaque deposition remains a matter of debate. To address the possibility that a pre-plaque inflammatory process occurs, we investigated the status of neuronal, astrocytic, and microglial markers in pre- and post-amyloid plaque stages in a novel transgenic rat model of an AD-like amyloid pathology (McGill-R-Thy1-APP). In this model, we found a marked upregulation of several classical inflammatory markers such as COX-2, IL-1β, TNF-α, and fractalkine (CX3CL1) in the cerebral cortex and hippocampus. Interestingly, many of these markers were highly expressed in amyloid beta-burdened neurons. Activated astrocytes and microglia were associated with these Aβ-burdened neurons. These findings confirm the occurrence of a proinflammatory process preceding amyloid plaque deposition and suggest that Aβ-burdened neurons play a crucial role in initiating inflammation in AD.
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%