Merck
  • Asiatic acid ameliorates dextran sulfate sodium-induced murine experimental colitis via suppressing mitochondria-mediated NLRP3 inflammasome activation.
Asiatic acid ameliorates dextran sulfate sodium-induced murine experimental colitis via suppressing mitochondria-mediated NLRP3 inflammasome activation.
International immunopharmacology (2014-12-20)
Wenjie Guo, Wen Liu, Biao Jin, Ji Geng, Jing Li, Hongqun Ding, Xuefeng Wu, Qiang Xu, Yang Sun, Jing Gao
ABSTRACT
In the present study, the effect of asiatic acid, a natural triterpenoid compound, on murine experimental colitis induced by dextran sulfate sodium (DSS) and its possible mechanism were examined in vivo and vitro. Oral administration of asiatic acid dose-dependently attenuated the loss of body weight and shortening of colon length induced by DSS. The disease activity index, histopathologic scores of musco and myeloperoxidase activity were also significantly reduced by asiatic acid treatment. Protein and mRNA levels of DSS-induced pro-inflammatory cytokines in colon, including TNF-α, IL-1β, IL-6 and IFN-γ, were markedly suppressed by asiatic acid. At the same time, decreased activation of caspase-1 in peritoneal macrophages was detected in asiatic acid-treated mice, which suggested that the NLRP3 inflammasome activation was suppressed. In addition, we also found that asiatic acid dose-dependently inhibited IL-1β secretion, caspase-1 activation as well as inflammasome assembling in vitro. Furthermore, the mechanism of asiatic acid was related to the inhibition of mitochondrial reactive oxygen species generation and prevention of mitochondrial membrane potential collapse. Taken together, our results demonstrate the ability of asiatic acid to inhibit NLRP3 inflammasome activation and its potential usage in the treatment of inflammatory bowel diseases.
Sigma-Aldrich
Sodium sulfate, JIS special grade, ≥99.0%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Asiatic acid, 97%
Sigma-Aldrich
Asiatic acid, ≥98% (HPLC), from Centella asiatica
Supelco
Asiatic acid, analytical standard