Merck
  • Pathologic staging of white matter lesions in adult-onset leukoencephalopathy/leukodystrophy with axonal spheroids.
Pathologic staging of white matter lesions in adult-onset leukoencephalopathy/leukodystrophy with axonal spheroids.
Journal of neuropathology and experimental neurology (2015-02-11)
Murad Alturkustani, Julia Keith, Lili-Naz Hazrati, Rosa Rademakers, Lee-Cyn Ang
ABSTRACT
The pathologic features of adult-onset leukoencephalopathy/leukodystrophy with axonal spheroids (ALAS) are variable, and this has led to different hypotheses as to whether primarily demyelination or axonopathy may underlie this disorder. Typical ALAS pathology is rarely accompanied by focal multiple sclerosis (MS)-like plaques. In ALAS pathology accompanied by focal multiple sclerosis (MS)-like plaques cases, the pathologic features cannot be distinguished from those of progressive MS with diffusely abnormal white matter. To clarify these issues, we examined neuropathologic features in 159 representative samples from 5 ALAS cases (3 men and 2 women aged 39-61 years) and in 95 representative samples from 3 chronic MS cases (1 man and 2 women aged 50-73 years). The white matter abnormalities in ALAS cases were characterized by 3 evolving stages: 1) white matter with numerous spheroids in a background of well-myelinated fibers; 2) moderate loss of myelinated fibers with sparse to moderate number of spheroids; and 3) leukodystrophy-like pattern of confluent axonal and myelin loss. The application of this staging system suggests that myelin loss in ALAS is preceded by axonopathy. In progressive MS cases, the diffusely abnormal white matter pathology could be attributed to both primary demyelination and axonopathy. Some cases with predominant axonopathy are difficult to distinguish from cases with ALAS.
Sigma-Aldrich
Periodic acid, 99.999% trace metals basis
Sigma-Aldrich
4-Aminosalicylic acid, 99%
Sigma-Aldrich
Periodic acid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Periodic acid, SAJ special grade, ≥98.5%
Sigma-Aldrich
Periodic acid, ACS reagent, 99%
Sigma-Aldrich
Periodic acid, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Periodic acid, puriss. p.a., ACS reagent, for oxidimetric titration, crystallized, ≥99.0% (RT)