Merck
  • Serum immunoglobulin free light chain levels are higher in girls than boys during eosinophilic oesophagitis.
Serum immunoglobulin free light chain levels are higher in girls than boys during eosinophilic oesophagitis.
Acta paediatrica (Oslo, Norway : 1992) (2014-04-05)
Karen Knipping, Diana Colson, Pascale Soulaines, Frank Redegeld, Johan Garssen, Christophe Dupont
ABSTRACT
Eosinophilic oesophagitis (EO) is an emerging worldwide disease, closely associated with male gender and allergic disorders. This study investigated the distribution of allergy markers in a cohort of children with EO. We analysed allergy markers in 91 children (62 males and 29 females) with EO and a control group of 45 age-matched children who had non-EO gastrointestinal allergic symptoms. The markers analysed were serum cow's milk-specific and hen's egg-specific IgE, thymic stromal lymphopoietin (TSLP), thymus-regulated and activation-regulated chemokine (TARC/CCL17) and immunoglobulin free light chain (Ig-fLC). In the EO group, cow's milk-specific IgE levels were detectable in 41.9% of males and 62.1% of females and hen's egg-specific levels in 25% of males and 26.9% of females. There was no gender difference in increased TSLP or TARC levels. Kappa Ig-fLC were increased in 5.6% of males and 20.8% of females (p = 0.058) and lambda Ig-fLC in 1.9% of males and 33.3% of females (p = 0.000). No gender differences were found in the control group. Our findings suggest that serum TSLP might be a potential marker of EO and TARC of non-EO gastrointestinal food allergies. In EO, serum Ig-fLC appeared higher in females, adding another gender difference to the biology of EO.
Sigma-Aldrich
Sulfuric acid solution, 0.1 M
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Sigma-Aldrich
Sulfuric acid solution, 5 mM
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Sigma-Aldrich
Sulfuric acid solution, 0.025 M
Sigma-Aldrich
Sulfuric acid solution, 1.5 M
Sigma-Aldrich
Sulfuric acid solution, 0.01 M
Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid solution, 0.5 M
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Human TARC / CCL17 ELISA Kit, for serum, plasma, cell culture supernatant and urine