Merck
  • Selective and strain-specific NFAT4 activation by the Toxoplasma gondii polymorphic dense granule protein GRA6.
Selective and strain-specific NFAT4 activation by the Toxoplasma gondii polymorphic dense granule protein GRA6.
The Journal of experimental medicine (2014-09-17)
Ji Su Ma, Miwa Sasai, Jun Ohshima, Youngae Lee, Hironori Bando, Kiyoshi Takeda, Masahiro Yamamoto
ABSTRACT
Toxoplasma gondii infection results in co-option and subversion of host cellular signaling pathways. This process involves discharge of T. gondii effector molecules from parasite secretory organelles such as rhoptries and dense granules. We report that the T. gondii polymorphic dense granule protein GRA6 regulates activation of the host transcription factor nuclear factor of activated T cells 4 (NFAT4). GRA6 overexpression robustly and selectively activated NFAT4 via calcium modulating ligand (CAMLG). Infection with wild-type (WT) but not GRA6-deficient parasites induced NFAT4 activation. Moreover, GRA6-deficient parasites failed to exhibit full virulence in local infection, and the treatment of WT mice with an NFAT inhibitor mitigated virulence of WT parasites. Notably, NFAT4-deficient mice displayed prolonged survival, decreased recruitment of CD11b(+) Ly6G(+) cells to the site of infection, and impaired expression of chemokines such as Cxcl2 and Ccl2. In addition, infection with type I parasites culminated in significantly higher NFAT4 activation than type II parasites due to a polymorphism in the C terminus of GRA6. Collectively, our data suggest that GRA6-dependent NFAT4 activation is required for T. gondii manipulation of host immune responses to maximize the parasite virulence in a strain-dependent manner.
Product Description
Isoflurane, European Pharmacopoeia (EP) Reference Standard
USP
Pyrimethamine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Xanthine, BioUltra, ≥99%
Sigma-Aldrich
Xanthine, ≥99.5% (HPLC), purified by recrystallization
Sigma-Aldrich
Xanthine, ≥99%
Pyrimethamine, European Pharmacopoeia (EP) Reference Standard
Supelco
Pyrimethamine, VETRANAL®, analytical standard
Sigma-Aldrich
DAPI, for nucleic acid staining