Merck
  • Souroubea sympetala (Marcgraviaceae): a medicinal plant that exerts anxiolysis through interaction with the GABAA benzodiazepine receptor.
Souroubea sympetala (Marcgraviaceae): a medicinal plant that exerts anxiolysis through interaction with the GABAA benzodiazepine receptor.
Canadian journal of physiology and pharmacology (2014-08-21)
Martha Mullally, Christian Cayer, Kari Kramp, Marco Otárola Rojas, Pablo Sanchez Vindas, Mario Garcia, Luis Poveda Alvarez, Tony Durst, Zul Merali, Vance L Trudeau, John T Arnason
ABSTRACT
The mode of action of the anxiolytic medicinal plant Souroubea sympetala was investigated to test the hypothesis that extracts and the active principle act at the pharmacologically important GABAA-benzodiazepine (GABAA-BZD) receptor. Leaf extracts prepared by ethyl acetate extraction or supercritical extraction, previously determined to have 5.54 mg/g and 6.78 mg/g of the active principle, betulinic acid, respectively, reduced behavioural parameters associated with anxiety in a rat model. When animals were pretreated with the GABAA-BZD receptor antagonist flumazenil, followed by the plant extracts, or a more soluble derivative of the active principle, the methyl ester of betulinic acid (MeBA), flumazenil eliminated the anxiety-reducing effect of plant extracts and MeBA, demonstrating that S. sympetala acts via an agonist action on the GABAA-BZD receptor. An in vitro GABAA-BZD competitive receptor binding assay also demonstrated that S. sympetala extracts have an affinity for the GABAA-BZD receptor, with an EC50 value of 123 μg/mL (EtOAc leaf extract) and 154 μg/mL (supercritical CO2 extract). These experiments indicate that S. sympetala acts at the GABAA-BZD receptor to elicit anxiolysis.
Sigma-Aldrich
Betulinic acid, ≥98% (HPLC)
Alfadex, European Pharmacopoeia (EP) Reference Standard
Supelco
Betulinic acid, analytical standard
Sigma-Aldrich
Betulinic acid, technical grade, 90%
Sigma-Aldrich
α-Cyclodextrin, Produced by Wacker Chemie AG, Burghausen, Germany, Life Science, 98.0-101.0% cyclodextrin basis (HPLC)
Sigma-Aldrich
Ethyl acetate, suitable for HPLC
Sigma-Aldrich
Ethyl acetate, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethyl acetate, JIS special grade, ≥99.5%
USP
Alpha Cyclodextrin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
α-Cyclodextrin, purum, ≥98.0% (HPLC)
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Flumazenil, >99% (HPLC), solid
Sigma-Aldrich
α-Cyclodextrin, ≥98%
Sigma-Aldrich
α-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
α-Cyclodextrin, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0% (HPLC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ethyl acetate, JIS 1000, ≥99.5%, for residue analysis
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate