Merck
  • Osmotically controlled pulsatile release capsule of montelukast sodium for chronotherapy: statistical optimization, in vitro and in vivo evaluation.
Osmotically controlled pulsatile release capsule of montelukast sodium for chronotherapy: statistical optimization, in vitro and in vivo evaluation.
Drug delivery (2013-11-13)
Om Prakash Ranjan, Usha Y Nayak, M Sreenivasa Reddy, Swapnil J Dengale, Prashant B Musmade, Nayanabhirama Udupa
ABSTRACT
The purpose of present study was to design, optimize and evaluate osmotically controlled pulsatile release capsule (PRC) of montelukast sodium (MKS) for the prevention of episodic attack of asthma in early morning and associated allergic rhinitis. Assembly of the capsular systems consisted of push, active and plug tablet arranged from bottom to top in hard gelatin capsule. The capsule system was coated with a semi-permeable membrane of cellulose acetate and drilled towards plug side in cap. A three-factor, three-level central composite design (CCD) with α = 1 was introduced to execute the experiments and quadratic polynomial model was generated to predict and assess the independent variables with respect to the dependent variables. The composition of optimal formulation was determined as weight of push tablet 138 mg (coded value: +0.59), plug tablet 60 mg (coded value: +0.49) and coating weight gain of 8.4 mg (coded value: -0.82). The results showed that the optimal formulation of PRCs had lag time of 4.5 h, release at 6 and 12 h are 61.95% and 96.29%, respectively. The X-ray radiographic imaging study was carried out to monitor the in vivo behavior of developed barium sulfate-loaded PRCs in rabbits under fasting conditions. In vivo pharmacokinetic study revealed Tmax of 2 h for marketed tablets; however 7 h for PRCs with initial lag time of 4 h. Thus designed capsular system may be helpful for patients with episodic attack of asthma in early morning and associated allergic rhinitis.
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Montelukast Sodium, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Montelukast sodium, United States Pharmacopeia (USP) Reference Standard
Montelukast sodium, European Pharmacopoeia (EP) Reference Standard