Merck
  • Synergistic effect of pendant hydroxypropyl and pyrrolidine moieties randomly distributed along polymethacrylamide backbones on in vitro DNA-transfection.
Synergistic effect of pendant hydroxypropyl and pyrrolidine moieties randomly distributed along polymethacrylamide backbones on in vitro DNA-transfection.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-12-03)
J A Redondo, D Velasco, M Pérez-Perrino, H Reinecke, A Gallardo, A Pandit, C Elvira
ABSTRACT
In this work, the cationic monomer N-ethyl pyrrolidine methacrylamide (EPA) was copolymerized with the neutral monomer N-hydroxypropyl methacrylamide (HPMA) at different molar ratios obtaining linear random copolymers that were characterized and evaluated in vitro as non-viral gene carriers using murine Swiss 3T3 fibroblasts. The copolymers with excess or equimolar amount of EPA were able to complex DNA forming stable polyplexes with an average size between 50 and 200 nm, while the copolymers with an excess of HPMA do not. Cell viability was shown to depend on the EPA/HPMA molar ratio, exhibiting the equimolar copolymer poly (EPA-co-HPMA) 50:50 (EPA50) a full cytocompatibility, similar to the HPMA-rich systems. This copolymer EPA50 has also shown significantly higher transfection levels than the systems with other compositions and the positive controls poly L-lysine (PLL) and poly EPA (pEPA). This statistical equimolar copolymer EPA50 has unique properties related to its composition and microstructure, which allows it to complex DNA, showing an excellent biocompatibility and high transfection efficiency.
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
Sigma-Aldrich
1,1,2,2-Tetrabromoethane, 98%
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
1,1,2,2-Tetrabromoethane, purum p.a., for separation (of mineral compounds), ≥98.0% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile) solution, 0.2 M in toluene
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
1,1,2,2-Tetrabromoethane, SAJ first grade, ≥98.0%
Sigma-Aldrich
Ethidium bromide solution, 10 mg/mL
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), SAJ first grade, ≥98.0%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), recrystallized from methanol, 99%
Supelco
Triethylamine, analytical standard
Sigma-Aldrich
Ethidium bromide, ≥95.0%
Sigma-Aldrich
Amino-2-propanol, 93%
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Supelco
(±)-1-Amino-2-propanol, analytical standard