Merck
  • RANKL expression in normal and malignant breast tissue responds to progesterone and is up-regulated during the luteal phase.
RANKL expression in normal and malignant breast tissue responds to progesterone and is up-regulated during the luteal phase.
Breast cancer research and treatment (2014-07-11)
Hong Hu, Jun Wang, Akash Gupta, Ali Shidfar, Daniel Branstetter, Oukseub Lee, David Ivancic, Megan Sullivan, Robert T Chatterton, William C Dougall, Seema A Khan
ABSTRACT
The receptor activator of nuclear factor-κB ligand (RANKL) acts as a paracrine factor in progesterone-induced mammary epithelial proliferation and tumorigenesis. This evidence comes mainly from mouse models. Our aim was to examine whether RANKL expression in human normal and malignant breast is under the control of progesterone throughout the menstrual cycle. Breast epithelial samples were obtained by random fine needle aspiration (rFNA) of the contralateral unaffected breasts (CUB) of 18 breast cancer patients, with simultaneous serum hormone measurements. Genes correlated with serum progesterone levels were identified through Illumina microarray analysis. Validation was performed using qRT-PCR in rFNA samples from CUB of an additional 53 women and using immunohistochemistry in tissue microarrays of 61 breast cancer samples. Expression of RANKL, DIO2, and MYBPC1 was correlated with serum progesterone in CUB, and was significantly higher in luteal phase. RANKL and MYBPC1 mRNA expression were highly correlated between CUB and matched tumor samples. RANKL protein expression was also significantly increased in the luteal phase and highly correlated with serum progesterone levels in cancer samples, especially in hormone receptor positive tumors. The regulatory effects of progesterone on the expression of RANKL, DIO2, and MYBPC1 were confirmed in three-dimensional cultures of normal breast organoids. In normal breast and in breast cancer, RANKL mRNA and protein expression fluctuate with serum progesterone with highest levels in the luteal phase, suggesting that RANKL is a modulator of progesterone signaling in normal and malignant breast tissue and a potential biomarker of progesterone action and blockade.
Product Description
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Progesterone, meets USP testing specifications
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Progesterone for system suitability, European Pharmacopoeia (EP) Reference Standard
Progesterone for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Progesterone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
tert-Butyl acetoacetate, reagent grade, 98%
Sigma-Aldrich
sRANKL human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
RANK Ligand/TRANCE human, >90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
MISSION® esiRNA, targeting human TNFSF11