Merck
  • Tuning the basicity of cyano-containing ionic liquids to improve SO2 capture through cyano-sulfur interactions.
Tuning the basicity of cyano-containing ionic liquids to improve SO2 capture through cyano-sulfur interactions.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-02-18)
Guokai Cui, Fengtao Zhang, Xiuyuan Zhou, Haoran Li, Jianji Wang, Congmin Wang
ABSTRACT
A new approach has been developed to improve SO2 sorption by cyano-containing ionic liquids (ILs) through tuning the basicity of ILs and cyano-sulfur interaction. Several kinds of cyano-containing ILs with different basicity were designed, prepared, and used for SO2 capture. The interaction between these cyano-containing ILs and SO2 was investigated by FTIR and NMR methods. Spectroscopic investigations and quantum chemical calculations showed that dramatic effects on SO2 capacity originate from the basicity of the ILs and enhanced cyano-sulfur interaction. Furthermore, the captured SO2 was easy to release by heating or bubbling N2 through the ILs. This efficient and reversible process, achieved by tuning the basicity of ILs, is an excellent alternative to current technologies for SO2 capture.
Sigma-Aldrich
Pyrrole-2-carbonitrile, 96%
Sigma-Aldrich
4-Cyanophenol, 95%
Sigma-Aldrich
Sodium cyanate, 96%
Sigma-Aldrich
Trihexyltetradecylphosphonium bromide, ≥95%
Sigma-Aldrich
4-Cyanobenzoic acid, 99%
Sigma-Aldrich
1,3-Propylene sulfite, 99%