Merck
SnAP reagents for the synthesis of piperazines and morpholines.
Organic letters (2014-02-08)
Michael U Luescher, Cam-Van T Vo, Jeffrey W Bode
ABSTRACT
Substituted piperazines and morpholines are valuable structural motifs in biologically active compounds, but are not easily prepared by contemporary cross-coupling approaches. In this report, we introduce SnAP reagents for the transformation of aldehydes into N-unprotected piperazines and morpholines. This approach offers simple, mild conditions compatible with aromatic, heteroaromatic, aliphatic, and glyoxylic aldehydes and provides mono- and disubstituted N-heterocycles in a single step.
Sigma-Aldrich
SnAP OA Reagent
Sigma-Aldrich
SnAP Pip Reagent
Sigma-Aldrich
SnAP DA Reagent
Sigma-Aldrich
SnAP M Reagent, 95%
Sigma-Aldrich
SnAP 3Me-M Reagent