Merck
Synthesis of mesoporous silica nanoparticles.
Chemical Society reviews (2013-02-14)
Si-Han Wu, Chung-Yuan Mou, Hong-Ping Lin
ABSTRACT
Good control of the morphology, particle size, uniformity and dispersity of mesoporous silica nanoparticles (MSNs) is of increasing importance to their use in catalyst, adsorption, polymer filler, optical devices, bio-imaging, drug delivery, and biomedical applications. This review discusses different synthesis methodologies to prepare well-dispersed MSNs and hollow silica nanoparticles (HSNs) with tunable dimensions ranging from a few to hundreds of nanometers of different mesostructures. The methods include fast self-assembly, soft and hard templating, a modified Stöber method, dissolving-reconstruction and modified aerogel approaches. In practical applications, the MSNs prepared by these methods demonstrate good potential for use in high-performance catalysis, antireflection coating, transparent polymer-MSNs nanocomposites, drug-release and theranostic systems.
Sigma-Aldrich
Silica, mesoporous, 200 nm particle size, pore size ~2 nm
Sigma-Aldrich
Silica, mesoporous SBA-15, <150 μm particle size, pore size 12 nm, Hexagonal pore morphology
Sigma-Aldrich
Silica, mesoporous, 0.5 μm particle size, pore size ~4 nm
Sigma-Aldrich
Silica, mesoporous, 2 μm particle size, pore size ~4 nm
Sigma-Aldrich
Silica, mesoporous, 3 μm particle size, pore size ~4 nm
Sigma-Aldrich
Silica, mesoporous SBA-15, <150 μm particle size, pore size 8 nm, Hexagonal pore morphology
Sigma-Aldrich
Silica, mesoporous, 1 μm particle size, pore size ~4 nm
Sigma-Aldrich
Silica, mesoporous, 1 μm particle size, pore size ~2 nm
Sigma-Aldrich
Silica, mesoporous SBA-15, <150 μm particle size, pore size 6 nm, Hexagonal pore morphology
Sigma-Aldrich
Silica, mesoporous SBA-15, <150 μm particle size, pore size 4 nm, Hexagonal pore morphology
Sigma-Aldrich
Silica, mesoporous SBA-16, <150 μm particle size, pore size 5 nm, Cubic pore morphology
Sigma-Aldrich
Silica, mesoporous, 2 μm particle size, pore size ~2 nm
Sigma-Aldrich
Silica, mesoporous, 3 μm particle size, pore size ~2 nm
Sigma-Aldrich
Silica, mesoporous MCM-48, ≤15 μm particle size, pore size 3 nm, Cubic pore morphology
Sigma-Aldrich
Silica mesoporous SBA-15, <150 μm particle size, pore size 10 nm, Hexagonal pore morphology