Merck
Formulation and evaluation of suspensions: mefenamic acid prodrugs.
Pakistan journal of pharmaceutical sciences (2014-07-13)
Kamal Shah, Sushant K Shrivastava, Pradeep Mishra
ABSTRACT
Gastrosparing novel prodrugs (MAM and MAT) synthesized consisted of mefenamic acid (MA) with menthol (M) and thymol (T). Structural characterizations of synthesized esters were done by Infra red spectroscopy (IR), proton nuclear magnetic resonance (1HNMR), mass spectroscopy. After evaluation of pharmacological i.e. anti-inflammatory, analgesic and ulcerogenic activities, the preformulation studies were undertaken. Based on these a few formulation (suspensions) were designed and prepared. The formulated suspensions were evaluated for content uniformity, sedimentation volume, recovery studies, redispersibility, viscosity, pH, particle size, zeta potential, effect of temperature and in-vitro dissolution rate. All the above parameters were found to be within the limit these indicated that the synthesized esters are good candidate for liquid dosage form. Thus it can be concluded synthesized prodrugs can be formulated in suspension form.
Product Description
Mefenamic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mefenamic acid
Supelco
Mefenamic acid, analytical standard