Merck
Propylene glycol dermatitis: re-evaluation of an old problem.
Contact dermatitis (1994-10-01)
J O Funk, H I Maibach
ABSTRACT
Evaluation of dermatitis associated with propylene glycol application or ingestion remains a challenge. The research dealing with skin reactions to propylene glycol is revisited and new aspects for future research are outlined. Based on literature review and our own observations, we propose classifying skin reactions to propylene glycol into 4 mechanisms: (a) irritant contact dermatitis, (b) allergic contact dermatitis, (c) non-immunologic contact urticaria, and (d) subjective or sensory irritation. This concept allows a partial explanation of effects observed by different authors. Despite attempts to define objective criteria, biologically, histopathologically, or clinically, the distinction between irritant and allergic reactions remains unclear. Furthermore, the irritation threshold of propylene glycol, and likewise the optimal standard concentration in patch tests, is sub judice. Future studies on propylene glycol dermatitis should include repeated patch tests with serial dose dilutions, repeated open application tests/provocative use tests, oral challenge tests, and biopsies for a more complete evaluation of mechanisms and clinical significance.
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
USP
Propylene glycol, United States Pharmacopeia (USP) Reference Standard
Supelco
1,2-Propanediol, analytical standard
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, SAJ special grade, ≥99.0%
Sigma-Aldrich
1,2-Propanediol, SAJ first grade, ≥99.0%
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%