Merck
  • Fluocinolone acetonide intravitreal implant for the treatment of diabetic macular edema.
Fluocinolone acetonide intravitreal implant for the treatment of diabetic macular edema.
Drug design, development and therapy (2013-06-06)
Wyatt B Messenger, Robert M Beardsley, Christina J Flaxel
ABSTRACT
Diabetic macular edema (DME) remains one of the leading causes of moderate to severe vision loss. Although laser photocoagulation was the standard of care for several years, few patients achieved significant improvements in visual acuity. As a result, several pharmacotherapies and surgeries have been investigated. The fluocinolone acetonide devices are one of the latest therapies considered for the treatment of DME. Despite bringing significant improvements in visual acuity, fluocinolone devices are associated with cataract formation, increased intraocular pressure (IOP), and surgery to lower IOP. Due to the risk of complications, fluocinolone acetonide devices should be considered only in cases refractive to first-line therapies. In this review, we evaluate current and emerging therapies for DME, with special emphasis on fluocinolone acetonide intravitreal devices.
Product Description
Fluocinolone acetonide, European Pharmacopoeia (EP) Reference Standard
USP
Fluocinolone acetonide, United States Pharmacopeia (USP) Reference Standard
Supelco
Fluocinolone acetonide, analytical standard
Supelco
Fluocinolone Acetonide, Pharmaceutical Secondary Standard; Certified Reference Material