Merck
Palladium-catalyzed oxidative carbonylation reactions.
ChemSusChem (2013-01-12)
Xiao-Feng Wu, Helfried Neumann, Matthias Beller
ABSTRACT
Palladium-catalyzed coupling reactions have become a powerful tool for advanced organic synthesis. This type of reaction is of significant value for the preparation of pharmaceuticals, agrochemicals, as well as advanced materials. Both, academic as well as industrial laboratories continuously investigate new applications of the different methodologies. Clearly, this area constitutes one of the major topics in homogeneous catalysis and organic synthesis. Among the different palladium-catalyzed coupling reactions, several carbonylations have been developed and widely used in organic syntheses and are even applied in the pharmaceutical industry on ton-scale. Furthermore, methodologies such as the carbonylative Suzuki and Sonogashira reactions allow for the preparation of interesting building blocks, which can be easily refined further on. Although carbonylative coupling reactions of aryl halides have been well established, palladium-catalyzed oxidative carbonylation reactions are also interesting. Compared with the reactions of aryl halides, oxidative carbonylation reactions offer an interesting pathway. The oxidative addition step could be potentially avoided in oxidative reactions, but only few reviews exist in this area. In this Minireview, we summarize the recent development in the oxidative carbonylation reactions.
Sigma-Aldrich
Palladium, powder, <75 μm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, 99.995% trace metals basis
Sigma-Aldrich
Palladium, powder or granules, 99.99% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Palladium, sponge, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Palladium, microfoil, disks, 25mm, thinness 0.25μm, specific density 303.4μg/cm2, permanent mylar 3.5μm support, 99.99%
Palladium, foil, not light tested, 50x50mm, thickness 0.005mm, 99.95%
Palladium, foil, light tested, 25x25mm, thickness 0.006mm, 99.95%
Palladium, rod, 10mm, diameter 6.0mm, 99.95%
Palladium, foil, not light tested, 50x50mm, thickness 0.008mm, 99.95%
Palladium, foil, not light tested, 25x25mm, thickness 0.0015mm, 99.95%
Palladium, wire reel, 1m, diameter 0.05mm, as drawn, 99.9%
Palladium, rod, 25mm, diameter 6.0mm, 99.95%
Palladium, wire reel, 0.2m, diameter 0.25mm, as drawn, 99.99+%
Palladium, wire reel, 0.05m, diameter 1.0mm, hard, 99.95%
Palladium, foil, not light tested, 100x100mm, thickness 0.008mm, 99.95%
Palladium, tube, 50mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, as drawn, 99.95%
Palladium, foil, not light tested, 50x50mm, thickness 0.00075mm, permanent polyester support, 99.75%
Palladium, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Palladium, foil, not light tested, 25x25mm, thickness 0.00075mm, permanent polyester support, 99.75%
Palladium, foil, not light tested, 25x25mm, thickness 0.006mm, 99.95%
Palladium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%
Palladium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.99+%
Palladium, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%