Merck
impact of aluminum exposure on the immune system: a mini review.
Environmental toxicology and pharmacology (2013-01-01)
Y Z Zhu, D W Liu, Z Y Liu, Y F Li
ABSTRACT
Aluminum (Al) is widely used in daily life and will lead to environmental release and exposure. The toxicity of Al had been documented, and which attracted a growing concern on human and animal health. The immune system appears to be sensitive to Al exposure. But few studies focused on the potential immunological responses induced by Al. It is imperative to study the effects of Al on the immune function and this review discusses the effects of Al on autoimmunity, oral tolerance, expression of the immune cells, hypersensitivity and erythrocyte immune function. It will provide evidence to study the association between Al and immune function.
Sigma-Aldrich
Aluminum, powder, <5 μm particle size, 99.5% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, ≤30 μm, spherical powder, ≥99%
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, powder, <75 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Aluminum, wire reel, 5m, diameter 2.0mm, as drawn, 99.999%
Aluminum, wire reel, 5m, diameter 0.25mm, as drawn, 99.999%
Aluminum, wire reel, 50m, diameter 0.25mm, hard, 99.99+%
Aluminum, wire reel, 5m, diameter 0.025mm, as drawn, 99.99+%
Aluminum, wire reel, 5m, diameter 0.025mm, as drawn, 99.0+%
Aluminum, honeycomb, 300x300mm, thickness 5mm, cell size 13mm, cell wall 0.1mm, core density 0.062g/cm3, phenolic adhesive (resin), 99.5%
Aluminum, foam, 150x150mm, thickness 6.35mm, bulk density 0.2g/cm3, porosity 0.93, 2 pores/cm, aluminum 6101 grade, 98.5%
Aluminum, honeycomb, 600x600mm, thickness 20mm, cell size 3.2mm, cell wall 0.025mm, core density 0.072g/cm3, phenolic adhesive (resin), 99.5%
Aluminum, honeycomb, 300x300mm, thickness 13.9mm, cell size 6.3mm, cell wall 0.064mm, core density 0.083g/cm3, phenolic adhesive (resin), facing skin aluminum 0.55mm, 99.9%
Aluminum, microleaf disks, 25mm, thinness 1.0μm, specific density 269.9μg/cm2, removable support, 99.999%
Aluminum, tube, 0.1m coil, as drawn, outside diameter 0.30mm, inside diameter 0.2mm, wall thickness 0.05mm, 99.5%
Aluminum, insulated wire, 20m, conductor diameter 0.1mm, insulation thickness 0.005mm, polyimide insulation, 99.95+%
Aluminum, rod, 100mm, diameter 20.0mm, as drawn, 99.999%
Aluminum, rod, 500mm, diameter 15.0mm, as drawn, 99.999%
Aluminum, rod, 50mm, diameter 28mm, as drawn, 99.999%
Aluminum, tube, 2m coil, as drawn, outside diameter 0.30mm, inside diameter 0.2mm, wall thickness 0.05mm, 99.5%
Aluminum, insulated wire, 2m, conductor diameter 0.1mm, insulation thickness 0.005mm, polyimide insulation, 99.95+%
Aluminum, tube, 1000mm, outside diameter 0.86mm, inside diameter 0.76mm, wall thickness 0.05mm, as drawn, 99.5%
Aluminum, sphere, 100pcs, diameter 2.5mm, aluminum alloy 2017 precision sphere, grade 200, 95.2%
Aluminum, rod, 200mm, diameter 15.0mm, as drawn, 99.999%
Aluminum, rod, 500mm, diameter 5.0mm, as drawn, 99.999%
Aluminum, rod, 200mm, diameter 20.0mm, as drawn, 99.999%