Merck
The use of oxidized zirconium alloy in knee arthroplasty.
Expert review of medical devices (2012-08-22)
Thomas J Heyse, Steven B Haas, Turgay Efe
ABSTRACT
Polyethylene wear is a key contributor to long-term failure in total knee arthroplasty. Oxidized zirconium alloy was created as a bearing surface for total joint arthroplasty in an attempt to address this concern. Oxidized zirconium has shown considerable improvements over existing materials in several key areas related to component longevity, including resistance to roughening, wear reduction, frictional behavior and biocompatibility. This review aims to summarize the current use of oxidized zirconium alloy in knee arthroplasty, as well as the possible advantages and downsides of this material.
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Zirconium, foil, 8mm disks, thickness 0.02mm, 99.2%
Zirconium, foil, light tested, 100x100mm, thickness 0.02mm, 99.2%
Zirconium, foil, 6mm disks, thickness 0.01mm, 99.2%
Zirconium, foil, 8mm disks, thickness 0.025mm, annealed, 99.2%
Zirconium, foil, 6mm disks, thickness 0.075mm, annealed, 99.2%
Zirconium, foil, light tested, 50x50mm, thickness 0.01mm, 99.2%
Zirconium, foil, light tested, 25x25mm, thickness 0.015mm, 99.2%
Zirconium, foil, light tested, 100x300mm, thickness 0.016mm, 99.2%
Zirconium, foil, light tested, 100x100mm, thickness 0.009mm, 99.2%
Zirconium, foil, not light tested, 50x50mm, thickness 0.004mm, 99.2%
Zirconium, foil, 6mm disks, thickness 0.5mm, annealed, 99.2%
Zirconium, foil, not light tested, 100x100mm, thickness 0.003mm, 99.2%
Zirconium, foil, 50mm disks, thickness 0.05mm, annealed, 99.2%
Zirconium, foil, 4mm disks, thickness 0.015mm, 99.2%
Zirconium, foil, 6mm disks, thickness 0.009mm, 99.2%
Zirconium, foil, light tested, 300x300mm, thickness 0.05mm, annealed, 99.2%
Zirconium, foil, not light tested, 100x100mm, thickness 0.006mm, 99.2%
Zirconium, foil, light tested, 100x100mm, thickness 0.01mm, 99.2%
Zirconium, foil, 8mm disks, thickness 0.016mm, 99.2%
Zirconium, foil, not light tested, 25x25mm, thickness 0.006mm, 99.2%
Zirconium, foil, light tested, 50x50mm, thickness 0.01mm, 99.2%
Zirconium, foil, light tested, 150x150mm, thickness 0.05mm, annealed, 99.2%
Zirconium, foil, not light tested, 150x150mm, thickness 0.008mm, 99.2%
Zirconium, foil, light tested, 25x25mm, thickness 0.009mm, 99.2%
Zirconium, rod, 100mm, diameter 12.7mm, centerless ground, 99.2%
Zirconium, rod, 1000mm, diameter 5.0mm, centerless ground, 99.2%