Merck
[Application of porous tantalum implant in treatment of bone defect].
Zhongguo xiu fu chong jian wai ke za zhi = Zhongguo xiufu chongjian waike zazhi = Chinese journal of reparative and reconstructive surgery (2012-03-13)
Jian Lu, Xiaoping Zheng, Zhiqiang Wang
ABSTRACT
To review the progress in the treatment of bone defect by porous tantalum implant. Recent literature was extensively reviewed and summarized, concerning the treatment method of bone defect by porous tantalum implant. By right of their unique properties, porous tantalum implants have achieved very good results in the treatment of certain types of bone defects. Porous tantalum implants have their own advantages and disadvantages. If the case is meet to its indications, this method can obtain a good effect. Porous tantalum implants provide a new way for the clinical treatment of bone defects.
Sigma-Aldrich
Tantalum, powder, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Tantalum, powder, 60-100 mesh, 99.9%
Tantalum, foil, 10mm disks, thickness 0.0125mm, 99.9%
Tantalum, foil, 10mm disks, thickness 0.015mm, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.01mm, 99.9%
Tantalum, foil, light tested, 150x150mm, thickness 0.03mm, as rolled, 99.9%
Tantalum, foil, light tested, 100x100mm, thickness 0.034mm, as rolled, 99.9%
Tantalum, foil, not light tested, 100x100mm, thickness 0.009mm, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.0005mm, 99.9%
Tantalum, rod, 100mm, diameter 6.35mm, annealed, 99.9%
Tantalum, foil, light tested, 100x100mm, thickness 0.03mm, as rolled, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.0015mm, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.075mm, annealed, 99.9%
Tantalum, tube, 1000mm, outside diameter 8.7mm, inside diameter 8.34mm, wall thickness 0.18mm, as drawn, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.02mm, 99.9%
Tantalum, rod, 50mm, diameter 12.7mm, annealed, 99.9%
Tantalum, tube, 100mm, outside diameter 2.1mm, inside diameter 1.6mm, wall thickness 0.25mm, as drawn, 99.9%
Tantalum, foil, light tested, 100x100mm, thickness 0.05mm, annealed, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.01mm, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.0015mm, 99.9%
Tantalum, pellets, 20g, max. size 10mm, 99.9%
Tantalum, tube, 100mm, outside diameter 0.31mm, inside diameter 0.19mm, wall thickness 0.06mm, as drawn, 99.9%
Tantalum, rod, 50mm, diameter 10.0mm, annealed, 99.9%
Tantalum, rod, 200mm, diameter 1.5mm, annealed, 99.9%
Tantalum, foil, not light tested, 82x100mm, thickness 0.006mm, 99.9%
Tantalum, tube, 100mm, outside diameter 3.0mm, inside diameter 2mm, wall thickness 0.5mm, as drawn, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.01mm, 99.9%
Tantalum, tube, 1000mm, outside diameter 1.05mm, inside diameter 0.65mm, wall thickness 0.2mm, as drawn, 99.9%
Tantalum, rod, 100mm, diameter 10.0mm, annealed, 99.9%