Merck
  • Occupational benzene exposure and the risk of lymphoma subtypes: a meta-analysis of cohort studies incorporating three study quality dimensions.
Occupational benzene exposure and the risk of lymphoma subtypes: a meta-analysis of cohort studies incorporating three study quality dimensions.
Environmental health perspectives (2010-10-01)
Jelle Vlaanderen, Qing Lan, Hans Kromhout, Nathaniel Rothman, Roel Vermeulen
ABSTRACT
The use of occupational cohort studies to assess the association of benzene and lymphoma is complicated by problems with exposure misclassification, outcome classification, and low statistical power. We performed meta-analyses of occupational cohort studies for five different lymphoma categories: Hodgkin lymphoma (HL), non-Hodgkin lymphoma (NHL), multiple myeloma (MM), acute lymphocytic leukemia (ALL), and chronic lymphocytic leukemia (CLL). We assessed three study quality dimensions to evaluate the impact of study quality variations on meta-relative risks (mRRs): stratification by the year of start of follow-up, stratification by the strength of the reported acute myelogenous leukemia association, and stratification by the quality of benzene exposure assessment. mRRs for MM, ALL, and CLL increased with increasing study quality, regardless of the study quality dimension. mRRs for NHL also increased with increasing study quality, although this effect was less pronounced. We observed no association between occupational benzene exposure and HL. Our meta-analysis provides support for an association between occupational benzene exposure and risk of MM, ALL, and CLL. The evidence for an association with NHL is less clear, but this is likely complicated by the etiologic heterogeneity of this group of diseases. Further consideration of the association between benzene and NHL will require delineation of risks by NHL subtype.
Sigma-Aldrich
Benzene, JIS special grade, ≥99.5%
Sigma-Aldrich
Benzene, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Benzene, SAJ first grade, ≥99.0%
Supelco
Benzene, analytical standard
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Supelco
Benzene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol