Merck
  • Analytical methodologies for the determination of omeprazole: an overview.
Analytical methodologies for the determination of omeprazole: an overview.
Journal of pharmaceutical and biomedical analysis (2007-05-29)
M Espinosa Bosch, A J Ruiz Sánchez, F Sánchez Rojas, C Bosch Ojeda
ABSTRACT
Omeprazole, a gastric acid pump inhibitor, dose-dependently controls gastric acid secretion; the drug has greater antisecretory activity than histamine H(2)-receptor antagonists. Omeprazole has been determined in formulations and biological fluids by a variety of methods such as spectrophotometry, high-performance liquid chromatography with ultraviolet detection and liquid chromatography coupled with tandem mass spectrometry. The overview includes the most relevant analytical methodologies used in its determination since the origin still today.
Product Description
Supelco
Omeprazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Omeprazole, solid
Omeprazole for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Omeprazole, United States Pharmacopeia (USP) Reference Standard
Supelco
Omeprazole, analytical standard