Merck
Fluorescence quenching of uranine on confeito-like Au nanoparticles.
Journal of nanoscience and nanotechnology (2014-04-25)
Masaki Ujihara, Nhut Minh Dang, Toyoko Imae
ABSTRACT
Effect of structure and size of Au nanoparticles (AuNPs) on fluorescence behavior of uranine was examined. Confeito-like AuNPs with different sizes (30 nm, 60 nm and 100 nm, respectively) had plasmon absorption bands at 555, 600 and 660 nm, while the band of spherical AuNP (20 nm in size) was at 525 nm. Fluorescence of uranine was significantly quenched by the small and medium confeito-like AuNPs, and the quenching effect by the large particle was less. In comparison, the spherical AuNP quenched more remarkable than the confeito-like AuNPs. A mechanism of resonance energy transfer from uranine to AuNPs via the surface plasmon was suggested, and the strong quenching effect of the small AuNPs could be explained by the energy transfer from adsorbed uranine molecules to AuNPs. These behaviors indicate that the large confeito-like AuNPs can be a preferable nano-probe and useful for plasmonic devices, which can tune or maintain the fluorescence properties of other markers.
Sigma-Aldrich
Gold, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, powder, <45 μm, 99.99% trace metals basis
Sigma-Aldrich
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Gold, insulated wire, 0.5m, conductor diameter 0.075mm, insulation thickness 0.012mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Gold, insulated wire, 0.5m, conductor diameter 0.125mm, insulation thickness 0.014mm, polyester insulation, 99.99%
Gold, microfoil, disks, 10mm, thinness 0.01μm, specific density 20.7μg/cm2, permanent mylar 3.5μm support, 99.99+%
Gold, insulated wire, 1m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyimide insulation, 99.99%
Gold, insulated wire, 0.5m, conductor diameter 0.125mm, insulation thickness 0.016mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Gold, insulated wire, 2m, conductor diameter 0.125mm, insulation thickness 0.014mm, polyester insulation, 99.99%
Gold, rod, 10mm, diameter 4.0mm, as drawn, 99.95%
Gold, insulated wire, 5m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyester insulation, 99.99%
Gold, microleaf disks, 10mm, thinness 0.5μm, specific density 966μg/cm2, removable support, 99.99+%
Gold, insulated wire, 0.5m, conductor diameter 0.025mm, insulation thickness 0.005mm, polyester insulation, 99.99%
Gold, tube, 200mm, outside diameter 1.0mm, inside diameter 0.5mm, wall thickness 0.25mm, as drawn, 99.95%
Gold, insulated wire, 1m, conductor diameter 0.05mm, insulation thickness 0.007mm, polyester insulation, 99.99%
Gold, rod, 6mm, diameter 6.0mm, as drawn, 99.95%
Gold, tube, 100mm, outside diameter 1.60mm, inside diameter 0.6mm, wall thickness 0.5mm, as drawn, 99.95%
Gold, rod, 10mm, diameter 6.0mm, as drawn, 99.95%
Gold, insulated wire, 2m, conductor diameter 0.075mm, insulation thickness 0.012mm, PTFE (polytetrafluoroethylene) insulation, 99.99%
Gold, microfoil, disks, 10mm, thinness 0.05μm, specific density 101.3μg/cm2, permanent mylar 3.5μm support, 99.99+%