Merck
Spectroscopic properties of Yb3+ doped CaNb2O6 phosphor.
Journal of nanoscience and nanotechnology (2014-04-18)
Ru Zhou, Xiantao Wei, Min Yin
ABSTRACT
The spectroscopic measurements of orthorhombic CaNb2O6 phosphor doped with Yb3+ ions have been performed. Low temperature spectra indicate that Yb3+ ions may occupy three types of nonequivalent sites inside the CaNb2O6 lattice: a main Yb3+ site dominating the near-infrared emission, and two other minor sites of similar behaviors to each other. An attempt to determine the Stark sublevels energies of the 2F7/2 and 2F5/2 manifolds of three Yb3+ nonequivalent ions was carried out. The fluorescence lifetimes associated to the emission lines of Yb3+ ions located at the main site and minor sites are also quite different. Furthermore, there exists energy transfer from the main site to the minor sites.
Sigma-Aldrich
Ytterbium, powder, ≥99.9% trace rare earth metals basis
Ytterbium, rod, 50mm, diameter 2.0mm, cast, 99%
Ytterbium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Ytterbium, rod, 100mm, diameter 6.35mm, cast, 99%
Ytterbium, rod, 50mm, diameter 10.0mm, cast, 99%
Ytterbium, lump, 25g, max. size 25mm, 99.9%
Ytterbium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Ytterbium, rod, 50mm, diameter 6.35mm, cast, 99%
Ytterbium, lump, 100g, max. size 25mm, 99.9%
Ytterbium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Ytterbium, lump, 50g, max. size 25mm, 99.9%
Sigma-Aldrich
Calcium oxide, 99.995% trace metals basis
Sigma-Aldrich
Calcium oxide, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium oxide, ReagentPlus®, 99.9% trace metals basis
Sigma-Aldrich
Calcium oxide, SAJ first grade, ≥98.0%
Sigma-Aldrich
Calcium oxide, nanopowder, <160 nm particle size (BET), 98%
Sigma-Aldrich
Calcium oxide, reagent grade