Merck
  • Pervasive nanoscale deformation twinning as a catalyst for efficient energy dissipation in a bioceramic armour.
Pervasive nanoscale deformation twinning as a catalyst for efficient energy dissipation in a bioceramic armour.
Nature materials (2014-04-01)
Ling Li, Christine Ortiz
ABSTRACT
Hierarchical composite materials design in biological exoskeletons achieves penetration resistance through a variety of energy-dissipating mechanisms while simultaneously balancing the need for damage localization to avoid compromising the mechanical integrity of the entire structure and to maintain multi-hit capability. Here, we show that the shell of the bivalve Placuna placenta (~99 wt% calcite), which possesses the unique optical property of ~80% total transmission of visible light, simultaneously achieves penetration resistance and deformation localization via increasing energy dissipation density (0.290 ± 0.072 nJ μm(-3)) by approximately an order of magnitude relative to single-crystal geological calcite (0.034 ± 0.013 nJ μm(-3)). P. placenta, which is composed of a layered assembly of elongated diamond-shaped calcite crystals, undergoes pervasive nanoscale deformation twinning (width ~50 nm) surrounding the penetration zone, which catalyses a series of additional inelastic energy dissipating mechanisms such as interfacial and intracrystalline nanocracking, viscoplastic stretching of interfacial organic material, and nanograin formation and reorientation.
Sigma-Aldrich
Marble, CP
USP
Calcium Carbonate (AS), United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Calcium carbonate, CP
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, ACS reagent, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Supelco
Calcium carbonate, Pharmaceutical Secondary Standard; Certified Reference Material