Merck
  • Ecotoxicity of engineered TiO2 nanoparticles to saltwater organisms: an overview.
Ecotoxicity of engineered TiO2 nanoparticles to saltwater organisms: an overview.
Environment international (2014-02-11)
D Minetto, G Libralato, A Volpi Ghirardini
ABSTRACT
The innovative properties of nanomaterials make them suitable for various applications in many fields. In particular, TiO2 nanoparticles (nTiO2) are widely used in paints, in cosmetics and in sunscreens that are products accessible to the mass market. Despite the great increase in the use of such nanomaterials, there is a paucity of general information about their potential effects to the aquatic species, especially to saltwater ones. Moreover, the difficulties of determining the effective exposure scenario make the acquired information low comparable. In this work, questions about the complexity of the real exposure scenario determination are discussed. The state of the art, concerning the experimental activities with nTiO2 toward the saltwater organisms is firstly illustrated, providing statistical information about the different matrices, organisms and nanoparticles employed. A comparison of the nTiO2 ecotoxicity effects, grouped by taxonomic classes, is provided illustrating their relative experimental conditions. Findings show the need to develop specific protocols for toxicity tests with ENPs to control the variability of experimental conditions. Some advices are finally proposed for the future experimental activities.
Sigma-Aldrich
Titanium(IV) oxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~10 nm × 10 μm
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~100 nm × 10 μm
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Titanium(IV) oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%
Sigma-Aldrich
Titanium(IV) oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis