Merck
  • Physicochemical properties of gelatin/silver nanoparticle antimicrobial composite films.
Physicochemical properties of gelatin/silver nanoparticle antimicrobial composite films.
Food chemistry (2013-11-23)
Paulraj Kanmani, Jong-Whan Rhim
ABSTRACT
Active nanocomposite films were prepared by blending aqueous solutions of gelatin with different concentrations of silver nanoparticles (AgNPs) using a solvent casting method. Formation of silver nanoparticles in the solution and films was confirmed with the surface plasmon resonance (SPR) band at 400-450 nm, measured by UV-vis absorption spectroscopy. The incorporation of AgNPs slightly affected the physical and mechanical properties of the films. Increase in the concentration of AgNPs resulted in a substantial decrease in water vapour permeability (WVP) and tensile strength (TS) of the gelatin films. Energy dispersive X-ray (EDX) spectroscopy and X-ray diffraction (XRD) analysis confirmed the presence of elemental silver and crystalline structure of the AgNPs in the gelatin film. Microscopic surface structure and thermal properties of the films were also examined by FE-SEM and thermogravimetric analysis. Gelatin/AgNPs nanocomposite films exhibited strong antibacterial activity against food-borne pathogens. Gelatin/AgNPs nanocomposite films are expected to have high potential as an active food packaging system to maintain food safety and to extend the shelf-life of packaged foods.
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver nanowires, diam. × L 50 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver nanowires, diam. × L 100 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Silver, conductive paste
Sigma-Aldrich
Silver, colloidal, 65-75% Ag basis
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Silver, foil, 10mm disks, thickness 0.15mm, as rolled, 99.95+%
Silver, foil, 10mm disks, thickness 0.125mm, as rolled, 99.99+%
Silver, foil, 0.5m coil, thickness 0.1mm, as rolled, 99.95+%
Silver, foil, 10mm disks, thickness 0.003mm, 99.9%
Silver, foil, 10mm disks, thickness 0.01mm, 99.95+%
Silver, foil, 10m coil, thickness 0.0125mm, coil width 9mm, 99.95+%
Silver, foil, 15mm disks, thickness 1.0mm, as rolled, 99.95+%
Silver, foil, 1m coil, thickness 0.05mm, annealed, 99.95+%
Silver, foil, 25mm disks, thickness 0.1mm, annealed, 99.95+%
Silver, foil, 25mm disks, thickness 1.0mm, as rolled, 99.99+%
Silver, foil, 4mm disks, thickness 0.004mm, 99.97%
Silver, foil, 1m coil, thickness 0.035mm, as rolled, 99.95+%
Silver, foil, 15mm disks, thickness 0.009mm, 99.97%
Silver, foil, 15mm disks, thickness 0.125mm, annealed, 99.95+%
Silver, foil, 4mm disks, thickness 0.1mm, as rolled, 99.95+%
Silver, foil, 1m coil, thickness 0.05mm, as rolled, 99.95+%
Silver, foil, 15mm disks, thickness 0.0125mm, 99.95+%
Silver, foil, 15mm disks, thickness 0.007mm, 99.97%
Silver, foil, 25mm disks, thickness 0.007mm, 99.97%
Silver, foil, 15mm disks, thickness 0.025mm, as rolled, 99.95+%
Silver, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.95+%