Merck
  • Multicenter, double-blind, parallel group study investigating the non-inferiority of efficacy and safety of a 2% miconazole nitrate shampoo in comparison with a 2% ketoconazole shampoo in the treatment of seborrhoeic dermatitis of the scalp.
Multicenter, double-blind, parallel group study investigating the non-inferiority of efficacy and safety of a 2% miconazole nitrate shampoo in comparison with a 2% ketoconazole shampoo in the treatment of seborrhoeic dermatitis of the scalp.
The Journal of dermatological treatment (2013-04-06)
Stanislaw A Buechner
ABSTRACT
This study investigated the non-inferiority of efficacy and tolerance of 2% miconazole nitrate shampoo in comparison with 2% ketoconazole shampoo in the treatment of scalp seborrheic dermatitis. A randomized, double-blind, comparative, parallel group, multicenter study was done. A total of 274 patients (145 miconazole, 129 ketoconazole) were enrolled. Treatment was twice-weekly for 4 weeks. Safety and efficacy assessments were made at baseline and at weeks 2 and 4. Assessments included symptoms of erythema, itching, scaling ['Symptom Scale of Seborrhoeic Dermatitis' (SSSD)], disease severity and global change [Clinical Global Impressions (CGIs) and Patient Global Impressions (PGIs)]. Miconazole shampoo is at least as effective and safe as ketoconazole shampoo in treating scalp seborrheic dermatitis scalp.
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
USP
Ketoconazole, United States Pharmacopeia (USP) Reference Standard
Ketoconazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Ketoconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Miconazole nitrate, European Pharmacopoeia (EP) Reference Standard
Miconazole, European Pharmacopoeia (EP) Reference Standard
USP
Miconazole nitrate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
(±)-Miconazole nitrate salt, imidazole antibiotic
Supelco
(±)-Miconazole nitrate salt, Pharmaceutical Secondary Standard; Certified Reference Material