Merck
  • Cilazapril stability in the presence of hydrochlorothiazide in model mixtures and fixed dose combination.
Cilazapril stability in the presence of hydrochlorothiazide in model mixtures and fixed dose combination.
Acta poloniae pharmaceutica (2014-01-05)
Sylwia K Paszun, Beata J Stanisz, Agnieszka Gradowska
ABSTRACT
The presented study aimed at the evaluation of hydrochlorothiazide influence on cilazapril stability in model mixture and fixed dose tablet formulation. The degradation of cilazapril in the presence of hydrochlorothiazide took place according to autocatalytic reaction kinetic mechanism, described mathematically by Prout-Tompkins equation. Hydrochlorothiazide coexistence with cilazapril in model mixture and fixed dose tablet without blister package accelerated cilazapril degradation in comparison with degradation of cilazapril substance. Values of reaction induction time shortened, while those of observed reaction rate constant increased. Increasing values of relative humidity and temperature have negative impact on cilazapril stability. Determined semi-logarithmic relationships: In k = f(RH) and Arrhenius ln k = f(1/T) are linear and are cilazapril stability predictive. The blister (OPA/Alu/PVC//Alu) package of fixed dose tablets, constitutes absolute moisture protection and prevent cilazapril--hydrochlorothiazide interaction occurrence.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Oxymetazoline hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Cilazapril impurity D, European Pharmacopoeia (EP) Reference Standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Oxymetazoline hydrochloride, ≥99%, solid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%