Merck
  • Treatment outcomes of injection laryngoplasty using cross-linked porcine collagen and hyaluronic acid.
Treatment outcomes of injection laryngoplasty using cross-linked porcine collagen and hyaluronic acid.
Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery (2013-10-17)
Ming-Hsun Wen, Po-Wen Cheng, Li-Jen Liao, Hsu-Wen Chou, Chi-Te Wang
ABSTRACT
To investigate the treatment outcomes and prognostic factors of injection laryngoplasty (IL) using cross-linked porcine collagen (PC) and hyaluronic acid (HA) in unilateral vocal fold paralysis (UVFP). Case series with chart review. A tertiary teaching hospital. This study reviewed 60 consecutive patients with UVFP who underwent IL with PC (n = 33) or HA (n = 27). Objective evaluations included maximal phonation time (MPT) and 10-item voice handicap index (VHI-10). Kaplan-Meier method was applied to evaluate the subjective treatment outcomes according to the patients' self-assessment of symptom recurrence via chart review for the follow-up period of 15 months. Log-rank tests were applied to evaluate the association between clinical factors and subjective treatment outcomes. Objective outcome measurements revealed significantly improved MPT and VHI-10 at 1, 3, and 6 months posttreatment, with nonsignificant differences between the PC and HA groups. Subjective treatment outcomes also revealed a nonsignificant difference between the 2 groups. The median symptom-free durations were 10.9 and 14.4 months for the PC and HA groups, respectively. Subsequent analyses failed to identify prognostic significance of sex, time to treatment, etiology, side, injection approaches, and the presence of aspiration. No significant adverse effects occurred during the follow-up period. This study demonstrated comparable subjective and objective improvements following IL using PC or HA in patients with UVFP. No significant prognostic factor of IL was discovered in the present research. Porcine collagen and HA as medium duration materials might play a role in the future of IL.
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 10,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 70,000-120,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 120,000-350,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 8,000-15,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 15,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 300,000-500,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 30,000-50,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,200
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,200,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 50,000-70,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,000,000-1,250,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,500,000-1,750,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,400,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus zooepidemicus, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from rooster comb, avian glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from bovine vitreous humor