Merck
  • The modality of cell-particle interactions drives the toxicity of nanosized CuO and TiO₂ in human alveolar epithelial cells.
The modality of cell-particle interactions drives the toxicity of nanosized CuO and TiO₂ in human alveolar epithelial cells.
Toxicology letters (2013-08-03)
Elisa Moschini, Maurizio Gualtieri, Miriam Colombo, Umberto Fascio, Marina Camatini, Paride Mantecca
ABSTRACT
Metal oxide NPs are abundantly produced in nanotech industries and are emitted in several combustion processes, suggesting the need to characterize their toxic impact on the human respiratory system. The acute toxicity and the morphological changes induced by copper oxide and titanium dioxide NPs (nCuO and nTiO₂) on the human alveolar cell line A549 are here investigated. Cell viability and oxidative stress have been studied in parallel with NP internalization and cell ultrastructural modifications. TiO₂ NPs were abundantly internalized by cells through the endocytic pathway, even they did not induce cell death and ultrastructural lesions. Only after 24h cells were affected by an abundant NP internalization presenting a consequent altered morphology. High cytotoxicity, oxidative stress and severe ultrastructural damages were produced by nCuO, since cell membrane and mitochondria resulted to be heavily affected, even at early exposure time. nCuO-induced toxicity has been interpreted as a consequence of both NPs reactivity and copper ions dissolution in lysosomal compartments, even the free NPs, scattered throughout all the cell compartments, might contribute to the toxicity. The antioxidant N-acetylcysteine was effective in recovering nCuO exposed cells viability and Bafilomycin A1 inhibited copper ions release in phagolysosomes and significantly rescued cells, suggesting a relevant cytotoxic mechanism relative to oxidative damages and authophagic cell death, together with NP internalization and dissolution. Our results support the previous data reporting CuO NPs are highly cytotoxic and genotoxic, and associate their toxic effects with their cell penetration and interaction with various compartments. In conclusion, the so-called "Trojan horse" mechanism and autophagy, are involved in nCuO-induced cell death, even a further research is needed to explain the events occurring at early exposure time.
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Copper(II) oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Copper(II) oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Copper(II) oxide, powder, 99.995% trace metals basis
Sigma-Aldrich
Copper(II) oxide, 99.999% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Copper(II) oxide, ACS reagent, ≥99.0%
Sigma-Aldrich
Copper(II) oxide on alumina, 14-30 mesh, extent of labeling: 13 wt. % loading
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Copper(II) oxide, needles, mixture of CuO and Cu2O, ACS reagent
Sigma-Aldrich
Copper(II) oxide, powder, <10 μm, 98%
Sigma-Aldrich
Titanium(IV) oxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Titanium(IV) oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Titanium(IV) oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%