Merck
  • Spatio-temporal distributions and the ecological and health risks of phthalate esters (PAEs) in the surface water of a large, shallow Chinese lake.
Spatio-temporal distributions and the ecological and health risks of phthalate esters (PAEs) in the surface water of a large, shallow Chinese lake.
The Science of the total environment (2013-06-19)
Wei He, Ning Qin, Xiangzhen Kong, Wenxiu Liu, Qishuang He, Huiling Ouyang, Chen Yang, Yujiao Jiang, Qingmei Wang, Bin Yang, Fuliu Xu
ABSTRACT
The spatio-temporal distributions and the ecological and health risks of PAEs in surface water of Lake Chaohu, the fifth largest lake in China, were studied based on the monthly monitoring of six PAE congeners from May 2010 to April 2011. The annual total concentration of the six PAE congeners (Σ6PAE) in the surface water ranged from 0.467 to 17.953 μg L(-1), with the average value of 4.042±3.929 μg L(-1). The di-n-butyl phthalate (DnBP) that dominated the Σ6PAE at 65.8% was found at its highest and lowest levels in the western lake (TX) and eastern drinking water source area (JC), respectively. The temporal distributions of Σ6PAE showed that the highest and lowest levels were observed in September 2010 and June 2010, respectively. The different relationships between the runoff and the PAEs with low and high levels of carbon might suggest their different sources. The DnBP had much greater ecological risks than the other studied PAE congeners as indicated by its potential affected fractions (PAFs) and the margin of safety (MOS10). The PAE congeners studied posed little health risk to the nearby male and female citizens.
Sigma-Aldrich
Potassium phthalate monobasic, JIS special grade, 99.8-100.2%
Supelco
Potassium phthalate monobasic, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Potassium phthalate monobasic, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
Phthalic acid dipotassium salt, 98%
Sigma-Aldrich
Potassium phthalate monobasic, ACS reagent, acidimetric standard
Supelco
Potassium phthalate monobasic, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Potassium phthalate monobasic, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Phthalic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Potassium hydrogen phthalate, BioXtra, ≥99.95%
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%