Merck
  • A general in situ hydrothermal rolling-up formation of one-dimensional, single-crystalline lead telluride nanostructures.
A general in situ hydrothermal rolling-up formation of one-dimensional, single-crystalline lead telluride nanostructures.
Small (Weinheim an der Bergstrasse, Germany) (2006-12-29)
Lizhi Zhang, Jimmy C Yu, Maosong Mo, Ling Wu, Kwan Wai Kwong, Quan Li
ABSTRACT
In this study we demonstrate that one-dimensional (1D) nanostructured lead telluride (PbTe) can be synthesized in a hydrothermal reaction between lead foil and tellurium powder. The resulting materials were characterized by X-ray diffraction, scanning electron microscopy, and transmission electron microscopy. The formation of the 1D structure can be explained by an in situ hydrothermal rolling-up mechanism whereby PbTe is formed hydrothermally and deposited on the lead substrate. The lead underneath the PbTe layer is then selectively etched by a cetyltrimethylammonium bromide solution, thus allowing the PbTe to roll up into 1D structures. This method can be extended to prepare other 1D tellurides, including CdTe, Cu(2)Te, and Ag(2)Te.
Product Description
Lead, foil, not light tested, 50x50mm, thickness 0.01mm, 99.99+%
Lead, foil, not light tested, 25x25mm, thickness 0.008mm, 99.99+%
Lead, foil, not light tested, 150x150mm, thickness 0.05mm, 99.99+%
Lead, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.99+%
Lead, foil, not light tested, 300x300mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, not light tested, 100x100mm, thickness 0.007mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.02mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.009mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.03mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, not light tested, 100x100mm, thickness 0.009mm, 99.99+%
Lead, foil, not light tested, 150x150mm, thickness 0.03mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.015mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.016mm, as rolled, 96% (Sn 1.5%, Sb 2.5%)
Lead, foil, not light tested, 100x100mm, thickness 0.04mm, 99.99+%
Lead, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, light tested, 25x25mm, thickness 0.03mm, 99.99+%
Lead, foil, not light tested, 50x50mm, thickness 0.03mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, not light tested, 50x50mm, thickness 0.015mm, 99.99+%
Lead, foil, 25mm disks, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 4mm disks, thickness 0.025mm, 99.99+%
Lead, foil, 4mm disks, thickness 0.009mm, 99.99+%
Lead, foil, 2m coil, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 4mm disks, thickness 0.015mm, 99.99+%
Lead, foil, 4mm disks, thickness 0.02mm, 99.99+%
Lead, foil, 25x25mm, thickness 1.0mm, as rolled, 99.5%
Lead, foil, 8mm disks, thickness 0.03mm, 99.99+%