Merck
  • Organic acids in the second morning urine in a healthy Swiss paediatric population.
Organic acids in the second morning urine in a healthy Swiss paediatric population.
Clinical chemistry and laboratory medicine (2004-01-08)
Olivier Boulat, Marianne Gradwohl, Vera Matos, Jean-Pierre Guignard, Claude Bachmann
ABSTRACT
Organic acid analysis is used for the early detection/ exclusion and for the follow-up of inherited disorders of amino acid and organic acid metabolism. Urinary organic acid concentrations in 417 healthy Caucasian children (1 day to 17 years of age) were determined after liquid solid extraction, as their trimethylsilyl derivatives, by gas chromatography and mass spectrometry. Concentrations of most of the organic acids adjusted for creatinine tend to decrease with age. No differences were found between gender except for the Krebs cycle intermediates in the older age groups. In neonates, the immaturity of the neonatal kidney led to a much larger variation of organic acid levels when related to creatinine. The low number of subjects (n = 36-52) per age class resulted in large 95% confidence intervals of the percentiles used for decision. This must be taken into account when using the data for exclusion or diagnosis of disorders.
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%
Sigma-Aldrich
Sarcosine, crystallized, ≥98.0% (T)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sarcosine, 98%
Sigma-Aldrich
cis-Aconitic acid, ≥98%
Sigma-Aldrich
Sarcosine, BioXtra
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
D-Erythronic acid potassium salt, ≥97.0% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenylpyruvic acid, 98%
Sigma-Aldrich
(S)-3-Hydroxybutyric acid, ≥97.0%
Sigma-Aldrich
mono-Ethyl malonate
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
Sigma-Aldrich
β-Hydroxyisovaleric acid, ≥95.0% (T)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)