Merck
  • Variation in enzymes of arylamine procarcinogen biotransformation among bladder cancer patients and control subjects.
Variation in enzymes of arylamine procarcinogen biotransformation among bladder cancer patients and control subjects.
Pharmacogenetics (2001-02-24)
S A Vaziri, N C Hughes, H Sampson, G Darlington, M A Jewett, D M Grant
ABSTRACT
Arylamines such as 2-naphthylamine and 4-aminobiphenyl are suspected human bladder procarcinogens that require bioactivation to DNA-reactive species to exert their carcinogenic potential. The goals of the present study were (i) to assay for the presence of the arylamine acetyltransferases NAT1 and NAT2, and of the cytochrome P450 isoform CYP1A2, in human bladder epithelium; and (ii) to determine whether the activities of these arylamine biotransforming enzymes differ between bladder cancer patients and control subjects. We measured in-vitro enzyme activities in biopsies of normal, undiseased bladder epithelium obtained from 103 bladder cancer patients. NAT1 activity was detectable in all samples, with mean levels higher than those found in human liver. Kinetic evidence also suggested low levels of NAT2 expression in this tissue, but there was no detectable CYP1A2 by either enzymatic or immunochemical measurements. We also compared several probe drug indices of in-vivo NAT1, NAT2 and CYP1A2 activity between 53 bladder cancer patients and 96 cancer-free control subjects who were carefully matched for age, gender and smoking status. NAT1 and NAT2 genotypes were also determined. No significant differences were found between bladder cancer patients and control subjects for a number of individual phenotypic or genotypic predictors of enzyme function. Our results suggest that although expression of particular arylamine biotransforming enzymes within the bladder tissue could play a significant role in locally bioactivating arylamine procarcinogens in theory, interindividual variations in CYP1A2, NAT1 and NAT2 activities do not significantly differ between bladder cancer patients and control subjects when potential arylamine exposures are controlled for
Product Description
Supelco
Aniline solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Aniline, SAJ first grade, ≥99.0%
Sigma-Aldrich
Aniline, JIS special grade, ≥99.0%
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Supelco
Aniline, analytical standard