Merck
  • β-Defensin 2 and 3 promote the uptake of self or CpG DNA, enhance IFN-α production by human plasmacytoid dendritic cells, and promote inflammation.
β-Defensin 2 and 3 promote the uptake of self or CpG DNA, enhance IFN-α production by human plasmacytoid dendritic cells, and promote inflammation.
Journal of immunology (Baltimore, Md. : 1950) (2013-06-19)
Poonam Tewary, Gonzalo de la Rosa, Neeraj Sharma, Luis G Rodriguez, Sergey G Tarasov, O M Zack Howard, Hidekazu Shirota, Folkert Steinhagen, Dennis M Klinman, De Yang, Joost J Oppenheim
ABSTRACT
Alarmins are a group of structurally diverse host defense antimicrobial peptides that are important immune activators. In this article, we present a novel role for two potent alarmins, human β-defensin 2 and 3 (HBD2 and 3), in promoting IFN-α production by human plasmacytoid dendritic cells. We demonstrate that HBD2 and 3 activate pDCs by enhancing the intracellular uptake of CpG and self DNA and promote DNA-induced IFN-α production in a TLR9-dependent manner. Both CpG and host DNA form aggregates that resemble DNA nets when combined with HBD2 and 3. Isothermal titration calorimetry studies to elucidate the nature of HBD3/CpG complexes demonstrate involvement of enthalpy-driven interactions, in addition to hydrophobic interactions, with the formation of complexes at a molar ratio of 2:1 defensin/CpG. The i.v. administration of HBD3/CpG complexes induced proinflammatory cytokines like IL-12, IFN-γ, IL-6, IFN-α, and IL-10 in serum, associated with an increased recruitment of APCs in the spleen. Subcutaneous injections of these complexes showed enhanced infiltration of inflammatory cells at the injection site, indicating a potential pathophysiological role for alarmin/DNA complexes in contributing to inflammation. Intraperitoneal immunization of HBD3/CpG complexes with OVA enhanced both cellular and humoral responses to OVA, compared with OVA/HBD3 or OVA/CPG alone, indicative of a much more potent adjuvant effect of the HBD3/CpG complexes. Thus, the ability of defensins to enhance cellular uptake of nucleic acids can lead to improved vaccine formulations by promoting their uptake by various cells, resulting in an enhanced immune response.
Product Description
Supelco
Albumin from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥90% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, powder, 62-88% (agarose gel electrophoresis)
Sigma-Aldrich
BD-3 human, recombinant, expressed in E. coli