Merck
Indoles synthesized from amines via copper catalysis.
Organic letters (2013-03-15)
Ronald Besandre, Miguel Jaimes, Jeremy A May
ABSTRACT
N-substituted indoles are synthesized from primary amines through a tandem reaction sequence. Initial condensation of the amine with an α-(o-haloaryl)ketone or aldehyde is followed by intramolecular aryl amination catalyzed by CuI. A variety of anilines and alkyl amines, including those with significant steric demands, are converted to indoles in high yields and with varying indole substitution.
Product Description
Supelco
Aniline solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Aniline-1-13C, 99 atom % 13C
Sigma-Aldrich
Aniline hydrochloride, ≥99%
Sigma-Aldrich
Aniline-4-13C, 99 atom % 13C
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Supelco
Aniline, analytical standard
Sigma-Aldrich
Aniline, JIS special grade, ≥99.0%
Sigma-Aldrich
Aniline, SAJ first grade, ≥99.0%